Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3559 |
Gene Name: | interleukin 2 receptor subunit alpha |
Gene Aliases: |
CD25, IDDM10, IL2R, IMD41, TCGFR, p55 |
Location: |
Chr.10:6010694-6062370 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IL2RA | NM_000417.2 | NP_000408.1 | ||
NM_001308242.1 | NP_001295171.1 | |||
NM_001308243.1 | NP_001295172.1 | |||
AK223313.1 | BAD97033.1 | |||
AK313847.1 | ||||
CD701954.1 | ||||
CR996090.1 | ||||
K03122.1 | AAB59535.1 | |||
X01057.1 | CAA25525.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1e49 | Chr.10:5684858 - 6183224 on Build GRCh38 | Loss | GDI2 IL2RA PFKFB3 RBM17 ANKRD16 IL15RA FBXO18 FAM208B MIR3155B MIR3155A |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |