Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 1813 |
Gene Name: | dopamine receptor D2 |
Gene Aliases: |
D2DR, D2R |
Location: |
Chr.11:113409595-113475279 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DRD2 | NM_000795.3 | NP_000786.1 | ||
NM_016574.3 | NP_057658.2 | |||
XM_017017296.1 | XP_016872785.1 | |||
AK308777.1 | ||||
BC021195.2 | AAH21195.1 | |||
BF437994.1 | ||||
DA693976.1 | ||||
M30625.1 | AAA88024.1 | |||
S69899.1 | AAB20571.1 | |||
X51362.1 | CAA35746.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv556444 | Chr.11:113318201 - 113512672 on Build GRCh38 | Gain | ANKK1 TTC12 DRD2 MIR4301 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |