Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3073 |
Gene Name: | hexosaminidase subunit alpha |
Gene Aliases: |
TSD |
Location: |
Chr.15:72343435-72376473 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HEXA | NM_000520.5 | NP_000511.2 | ||
NM_001318825.1 | NP_001305754.1 | |||
NR_134869.1 | ||||
AK222502.1 | BAD96222.1 | |||
AK296528.1 | ||||
AK297678.1 | ||||
AK301000.1 | ||||
AK301138.1 | ||||
AK307770.1 | ||||
AK310154.1 | ||||
BC018927.2 | AAH18927.1 | |||
BC084537.1 | AAH84537.1 | |||
BP356349.1 | ||||
CR627386.1 | CAH10482.1 | |||
DC356933.1 | ||||
KU178109.1 | ||||
KU178110.1 | ||||
KU178111.1 | ||||
KU178112.1 | ||||
KU178113.1 | ||||
M13520.1 | AAA51827.1 |
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |