Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 146057 |
Gene Name: | tau tubulin kinase 2 |
Gene Aliases: |
SCA11, TTBK |
Location: |
Chr.15:42738974-42920995 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TTBK2 | NM_173500.3 | NP_775771.3 | ||
XM_005254171.4 | XP_005254228.1 | |||
XM_005254173.4 | XP_005254230.1 | |||
XM_006720402.3 | XP_006720465.1 | |||
XM_006720403.3 | XP_006720466.1 | |||
XM_017021950.1 | XP_016877439.1 | |||
AF525400.1 | AAO14996.1 | |||
AK131372.1 | BAD18523.1 | |||
BC041876.1 | AAH41876.1 | |||
BC071556.1 | AAH71556.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525461 | Chr.15:42770350 - 43189929 on Build GRCh38 | Loss | UBR1 TTBK2 CCNDBP1 TMEM62 |
nsv528725 | Chr.15:42463357 - 42812208 on Build GRCh38 | Gain | HAUS2 ZNF106 TTBK2 CDAN1 STARD9 LRRC57 SNAP23 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |