Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3990 |
Gene Name: | lipase C, hepatic type |
Gene Aliases: |
HDLCQ12, HL, HTGL, LIPH |
Location: |
Chr.15:58410754-58568952 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPC | NM_000236.2 | NP_000227.2 | ||
XM_005254372.1 | XP_005254429.1 | |||
AK290062.1 | ||||
AK292631.1 | ||||
AK293324.1 | ||||
AK309448.1 | ||||
AK315306.1 | ||||
BC132825.1 | ||||
BC136495.1 | ||||
BC146659.1 | ||||
D83548.1 | BAA12014.1 | |||
J03540.1 | AAA59520.1 | |||
J03895.1 | AAA61165.1 | |||
X07228.1 | CAA30188.1 |
Entrez Gene ID: | 101928694 |
Gene Name: | LIPC antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:58434901-58498735 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPC-AS1 | NR_120338.1 | |||
AK091917.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv569600 | Chr.15:58439368 - 58486840 on Build GRCh38 | Loss | LIPC-AS1 LIPC |
nsv569598 | Chr.15:58260407 - 58560264 on Build GRCh38 | Loss | LIPC-AS1 LIPC LOC101928635 LOC101928668 |
nsv1043362 | Chr.15:58434138 - 58464372 on Build GRCh38 | Loss | LIPC-AS1 LIPC |
esv3581729 | Chr.15:58432325 - 58464372 on Build GRCh38 | Loss | LIPC-AS1 LIPC |
nsv1047048 | Chr.15:58409201 - 58492601 on Build GRCh38 | Loss | LIPC-AS1 LIPC LOC101928635 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |