Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 351 |
Gene Name: | amyloid beta precursor protein |
Gene Aliases: |
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2 |
Location: |
Chr.21:25880550-26171128 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APP | NM_000484.3 | NP_000475.1 | ||
NM_001136016.3 | NP_001129488.1 | |||
NM_001136129.2 | NP_001129601.1 | |||
NM_001136130.2 | NP_001129602.1 | |||
NM_001136131.2 | NP_001129603.1 | |||
NM_001204301.1 | NP_001191230.1 | |||
NM_001204302.1 | NP_001191231.1 | |||
NM_001204303.1 | NP_001191232.1 | |||
NM_201413.2 | NP_958816.1 | |||
NM_201414.2 | NP_958817.1 | |||
A02759.1 | CAA00268.1 | |||
AF282245.1 | AAQ14327.1 | |||
AK294534.1 | ||||
AK295621.1 | ||||
AK296229.1 | ||||
AK297229.1 | ||||
AK297412.1 | ||||
AK298861.1 | ||||
AK311717.1 | ||||
AK312326.1 | ||||
BC004369.1 | AAH04369.1 | |||
BC065529.1 | AAH65529.1 | |||
BI559391.1 | ||||
BM172428.1 | ||||
M28373.1 | AAA58727.1 | |||
X06989.1 | CAA30050.1 | |||
Y00264.1 | CAA68374.1 |
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |