Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 89781 |
Gene Name: | HPS4, biogenesis of lysosomal organelles complex 3 subunit 2 |
Gene Aliases: |
BLOC3S2, LE |
Location: |
Chr.22:26443613-26483863 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HPS4 | XM_011530492.1 | XP_011528794.1 | ||
XM_017029045.1 | XP_016884534.1 | |||
XM_017029048.1 | XP_016884537.1 | |||
XM_017029049.1 | XP_016884538.1 |
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |