Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2187 |
Gene Name: | Fanconi anemia complementation group B |
Gene Aliases: |
FA2, FAAP90, FAAP95, FAB, FACB |
Location: |
Chr.X:14690863-14873255 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FANCB | XM_011545470.2 | XP_011543772.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9934 | Chr.X:14730738 - 15173654 on Build GRCh38 | Loss | MOSPD2 FANCB GLRA2 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |