Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 4935 |
Gene Name: | G protein-coupled receptor 143 |
Gene Aliases: |
NYS6, OA1 |
Location: |
Chr.X:9725413-9765965 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR143 | NM_000273.2 | NP_000264.2 | ||
XM_005274541.3 | XP_005274598.1 | |||
BC068977.1 | AAH68977.1 | |||
Z48804.1 | CAA88742.1 |
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |