Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 8287 |
Gene Name: | ubiquitin specific peptidase 9, Y-linked |
Gene Aliases: |
DFFRY, SPGFY2 |
Location: |
Chr.Y:12701231-12860844 on Build GRCh38 |
Assay Gene Location: | Within Intron 36 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP9Y | NM_004654.3 | NP_004645.2 | ||
XM_017030078.1 | XP_016885567.1 | |||
AF000986.2 | AAC51833.1 | |||
Y13618.1 | CAA73940.1 | |||
Y13619.1 | CAA73941.1 |
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |