Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 6622 |
Gene Name: | synuclein alpha |
Gene Aliases: |
NACP, PARK1, PARK4, PD1 |
Location: |
Chr.4:89724099-89838324 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SNCA | NM_000345.3 | 6 | 2082 | NP_000336.1 |
NM_001146054.1 | 6 | 2078 | NP_001139526.1 | |
NM_001146055.1 | 6 | 1889 | NP_001139527.1 | |
NM_007308.2 | 5 | 1994 | NP_009292.1 | |
JN709859.1 | 5 | 2036 | ||
JN709860.1 | 4 | 1952 | ||
JN709861.1 | 5 | 1998 | ||
JN709862.1 | 5 | 2040 | ||
JN709863.1 | 4 | 1956 | ||
JN709864.1 | 5 | 1805 | ||
JN709865.1 | 5 | 1847 | ||
JN709866.1 | 4 | 1763 | ||
JN709867.1 | 5 | 1847 | ||
JN709868.1 | 5 | 1889 | ||
JN709869.1 | 4 | 1805 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1012406 | Chr.4:89246630 - 90245636 on Build GRCh38 | Loss | SNCA MMRN1 GPRIN3 SNCA-AS1 CCSER1 |
nsv594806 | Chr.4:88621130 - 90539537 on Build GRCh38 | Gain | SNCA MMRN1 HERC3 TIGD2 GPRIN3 NAP1L5 SNCA-AS1 CCSER1 FAM13A-AS1 FAM13A |
Set Membership: |
Intragenic Exonic DGV Variation |