Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 57654 |
Gene Name: | UV stimulated scaffold protein A |
Gene Aliases: |
KIAA1530, UVSS3 |
Location: |
Chr.4:1345662-1388049 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UVSSA | NM_001317934.1 | NP_001304863.1 | ||
NM_001317935.1 | NP_001304864.1 | |||
NM_020894.3 | NP_065945.2 | |||
XM_017008490.1 | XP_016863979.1 | |||
XM_017008491.1 | XP_016863980.1 | |||
XM_017008492.1 | XP_016863981.1 | |||
XM_017008493.1 | XP_016863982.1 | |||
XM_017008494.1 | XP_016863983.1 | |||
XM_017008495.1 | XP_016863984.1 | |||
XM_017008496.1 | 9 | 5398 | XP_016863985.1 | |
XM_017008498.1 | XP_016863987.1 | |||
AB040963.1 | ||||
BC110331.1 | AAI10332.1 | |||
BC140901.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | MAEA PDE6B FGFRL1 MYL5 NKX1-1 CTBP1-AS LOC100129917 LOC101928548 TMEM175 CPLX1 CRIPAK UVSSA CTBP1 LOC100130872 GAK MFSD7 SLC26A1 ATP5I LOC105374344 RNF212 SPON2 DGKQ LOC107986211 TMED11P CTBP1-AS2 LOC101928521 PCGF3 IDUA |
nsv593278 | Chr.4:1165892 - 1421910 on Build GRCh38 | Loss | CTBP1 LOC101928548 SPON2 MAEA LOC100130872 CTBP1-AS2 CRIPAK NKX1-1 UVSSA CTBP1-AS |
nsv593302 | Chr.4:1344830 - 1420896 on Build GRCh38 | Loss | CRIPAK NKX1-1 UVSSA |
Set Membership: |
Intragenic Exonic DGV Variation |