Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 8546 |
Gene Name: | adaptor related protein complex 3 beta 1 subunit |
Gene Aliases: |
ADTB3, ADTB3A, HPS, HPS2, PE |
Location: |
Chr.5:78002326-78294755 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AP3B1 | NM_001271769.1 | NP_001258698.1 | ||
NM_003664.4 | NP_003655.3 | |||
XM_005248618.3 | XP_005248675.1 | |||
XM_005248619.4 | XP_005248676.1 | |||
XM_017010001.1 | XP_016865490.1 | |||
AK225908.1 | ||||
AK291201.1 | ||||
AK303300.1 | ||||
AY623424.1 | AAT49048.1 | |||
BC038444.1 | AAH38444.1 | |||
BX538041.1 | CAD97982.1 | |||
KU178426.1 | ||||
KU178427.1 | ||||
KU178428.1 | ||||
U81504.1 | AAB61638.1 | |||
U91931.1 | AAD03778.1 |
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |