Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 7486 |
Gene Name: | Werner syndrome RecQ like helicase |
Gene Aliases: |
RECQ3, RECQL2, RECQL3 |
Location: |
Chr.8:31033262-31173761 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WRN | NM_000553.4 | NP_000544.2 | ||
XM_011544639.2 | XP_011542941.1 | |||
XM_011544640.1 | XP_011542942.1 | |||
AB209652.1 | BAD92889.1 | |||
AF091214.1 | AAC63361.1 | |||
AL833572.1 | ||||
AY818673.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831278 | Chr.8:31059468 - 31213390 on Build GRCh38 | Gain | WRN |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |