Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2260 |
Gene Name: | fibroblast growth factor receptor 1 |
Gene Aliases: |
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1 |
Location: |
Chr.8:38411138-38468834 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |