Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 8301 |
Gene Name: | phosphatidylinositol binding clathrin assembly protein |
Gene Aliases: |
CALM, CLTH, LAP |
Location: |
Chr.11:85957171-86069881 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1053808 | Chr.11:85877876 - 86046651 on Build GRCh38 | Gain | PICALM CCDC83 |
nsv1035489 | Chr.11:85791910 - 86067187 on Build GRCh38 | Gain | SYTL2 PICALM CCDC83 |
dgv2041n54 | Chr.11:85972511 - 86064017 on Build GRCh38 | Loss | PICALM |
dgv164n27 | Chr.11:85977166 - 86046547 on Build GRCh38 | Loss | PICALM |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |