Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 5781 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 11 |
Gene Aliases: |
BPTP3, CFC, JMML, METCDS, NS1, PTP-1D, PTP2C, SH-PTP2, SH-PTP3, SHP2 |
Location: |
Chr.12:112418732-112509913 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTPN11 | NM_002834.3 | NP_002825.3 | ||
NM_080601.1 | NP_542168.1 | |||
XM_006719526.2 | XP_006719589.1 | |||
XM_011538613.2 | XP_011536915.1 | |||
XM_017019722.1 | XP_016875211.1 | |||
AK289854.1 | ||||
BC008692.1 | AAH08692.1 | |||
BT007106.1 | AAP35770.1 | |||
D13540.1 | BAA02740.2 | |||
L03535.1 | AAA36611.1 | |||
L07527.1 | AAA17022.1 | |||
L08807.1 | ||||
X70766.1 | CAA50045.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv889 | Chr.12:112445258 - 112490108 on Build GRCh38 | Deletion | PTPN11 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |