Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 120892 |
Gene Name: | leucine rich repeat kinase 2 |
Gene Aliases: |
AURA17, DARDARIN, PARK8, RIPK7, ROCO2 |
Location: |
Chr.12:40224895-40369285 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRRK2 | NM_198578.3 | NP_940980.3 | ||
XM_005268629.3 | XP_005268686.1 | |||
XM_011537877.2 | XP_011536179.1 | |||
XM_011537881.2 | XP_011536183.1 | |||
XM_011537882.2 | XP_011536184.1 | |||
XM_017018786.1 | XP_016874275.1 | |||
XM_017018789.1 | XP_016874278.1 | |||
AK127729.1 | ||||
AY792511.1 | AAV63975.1 | |||
BC117180.1 | ||||
BM285390.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1048258 | Chr.12:39970608 - 41209299 on Build GRCh38 | Gain | CNTN1 SLC2A13 LRRK2 PDZRN4 LOC101930164 MUC19 |
nsv1042967 | Chr.12:40235834 - 40316517 on Build GRCh38 | Gain | LRRK2 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |