Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 55812 |
Gene Name: | spermatogenesis associated 7 |
Gene Aliases: |
HEL-S-296, HSD-3.1, HSD3, LCA3 |
Location: |
Chr.14:88385638-88470350 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |