Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 1311 |
Gene Name: | cartilage oligomeric matrix protein |
Gene Aliases: |
EDM1, EPD1, MED, PSACH, THBS5, TSP5 |
Location: |
Chr.19:18782773-18791305 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COMP | NM_000095.2 | NP_000086.2 | ||
AB086984.1 | BAC53888.1 | |||
AK074508.1 | BAC11031.1 | |||
AK223216.1 | BAD96936.1 | |||
AK290595.1 | ||||
AK296586.1 | ||||
AK297798.1 | ||||
AK297814.1 | ||||
BC110847.1 | AAI10848.1 | |||
BC125092.1 | ||||
L32137.1 | AAA57253.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion | TMEM161A KLHL26 DDX49 TM6SF2 LOC102724360 BORCS8 ARMC6 SLC25A42 CERS1 COPE SUGP1 NR2C2AP HAPLN4 COMP CRTC1 MEF2B RFXANK UPF1 GDF1 HOMER3 NCAN SUGP2 BORCS8-MEF2B |
nsv952422 | Chr.19:18606691 - 18807891 on Build GRCh38 | Deletion | CRLF1 KLHL26 COMP CRTC1 TMEM59L |
nsv833777 | Chr.19:18765126 - 18920267 on Build GRCh38 | Loss | COPE COMP DDX49 CRTC1 UPF1 GDF1 CERS1 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |