Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3172 |
Gene Name: | hepatocyte nuclear factor 4 alpha |
Gene Aliases: |
FRTS4, HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14 |
Location: |
Chr.20:44355801-44432845 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HNF4A | NM_001030003.2 | NP_001025174.1 | ||
NM_001030004.2 | NP_001025175.1 | |||
NM_001287182.1 | NP_001274111.1 | |||
NM_001287183.1 | NP_001274112.1 | |||
NM_001287184.1 | NP_001274113.1 | |||
NM_175914.4 | NP_787110.2 | |||
AY680696.1 | AAT91237.1 | |||
AY680697.1 | AAT91238.1 | |||
AY680698.1 | AAT91239.1 | |||
FJ608822.1 | ||||
HQ692868.1 |
Entrez Gene ID: | 101927219 |
Gene Name: | HNF4A antisense RNA 1 |
Gene Aliases: |
uc002xlx |
Location: |
Chr.20:44377743-44395706 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HNF4A-AS1 | NR_109949.1 | |||
AF143870.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1057847 | Chr.20:42652464 - 44431352 on Build GRCh38 | Loss | OSER1-AS1 L3MBTL1 HNF4A-AS1 LOC101927138 GTSF1L LOC105372626 MIR3646 MYBL2 OSER1 R3HDML LOC101927200 LOC101927159 TOX2 JPH2 SGK2 FITM2 IFT52 HNF4A GDAP1L1 PTPRT SRSF6 |
nsv586065 | Chr.20:44261867 - 44505244 on Build GRCh38 | Gain | LINC01430 HNF4A-AS1 R3HDML LINC01620 TTPAL FITM2 HNF4A MIR3646 GDAP1L1 SERINC3 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |