Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 14633 |
Gene Name: | GLI-Kruppel family member GLI2 |
Gene Aliases: |
AW546128 |
Location: |
Chr.1:118834061-119053619 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gli2 | NM_001081125.1 | 6 | 1087 | NP_001074594.1 |
AM261668.1 | 3 | 312 | ||
AM261669.1 | 3 | 331 | ||
AM261670.1 | 4 | 418 | ||
AM262741.1 | 4 | 418 | ||
AM262742.1 | 5 | 604 | ||
AM262743.1 | 5 | 701 | ||
AM262744.1 | 7 | 918 | ||
BC082604.1 | 6 | 1028 | ||
BC085190.1 | 6 | 1003 | ||
X99104.1 | 6 | 847 |
Set Membership: |
Intragenic Exonic |