Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 209176 |
Gene Name: | indoleamine 2,3-dioxygenase 2 |
Gene Aliases: |
AI265623, C230043N17Rik, Ido-2, Indol1 |
Location: |
Chr.8:24531892-24576404 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ido2 | NM_145949.2 | 3 | 345 | NP_666061.3 |
AK082385.1 | 2 | 422 | ||
BC026393.1 | 3 | 345 | AAH26393.1 | |
EF137182.1 | 3 | 345 | ||
EU440733.1 | 2 | 137 |
Set Membership: |
Intragenic Exonic |