Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 16197 |
Gene Name: | interleukin 7 receptor |
Gene Aliases: |
CD127, IL-7Ralpha |
Location: |
Chr.15:9506159-9529876 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il7r | NM_008372.4 | NP_032398.3 | ||
AF078906.1 | AAF06717.1 | |||
AK040364.1 | BAC30571.1 | |||
AK040740.1 | BAC30689.1 | |||
AK041838.1 | BAC31080.1 | |||
AK042264.1 | BAC31208.1 | |||
AK154215.1 | ||||
AK154516.1 | BAE32645.1 | |||
BC089571.1 | AAH89571.1 | |||
BY776757.1 | ||||
M29697.1 | AAA39304.1 |
Set Membership: |
Intragenic Non-exonic |