Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 17250 |
Gene Name: | ATP-binding cassette, sub-family C (CFTR/MRP), member 1 |
Gene Aliases: |
Abcc1a, Abcc1b, MRP, Mdrap, Mrp1 |
Location: |
Chr.16:14361521-14474879 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abcc1 | NM_008576.3 | 29 | 4232 | NP_032602.1 |
AF022908.1 | 29 | 4194 | AAB80938.1 | |
AF213389.1 | 2 | 232 | AAF31429.1 | |
AK029876.1 | 29 | 4233 | BAC26654.1 | |
AK045109.1 | BAC32226.1 | |||
AK049174.1 | 13 | 1999 | BAC33586.1 | |
AK140421.1 | 6 | 1775 | ||
BC090617.1 | 29 | 4203 |
Set Membership: |
Intragenic Exonic |