Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 17869 |
Gene Name: | myelocytomatosis oncogene |
Gene Aliases: |
AU016757, Myc2, Niard, Nird, bHLHe39 |
Location: |
Chr.15:61985341-61990361 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myc | NM_001177352.1 | NP_001170823.1 | ||
NM_001177353.1 | NP_001170824.1 | |||
NM_001177354.1 | NP_001170825.1 | |||
NM_010849.4 | NP_034979.3 | |||
AK087961.1 | BAC40060.1 | |||
AK133952.1 | BAE21948.1 | |||
AK145084.1 | BAE26228.1 | |||
BB858498.1 | ||||
BC138931.1 | ||||
BC138932.1 | ||||
X01023.1 | CAA25508.1 |
Set Membership: |
Intragenic Intronic Non-exonic Validated |