Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 001555
          Assay Name: hsa-miR-623
          miRBase Accession Number: MI0003637
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 99356131 - 99356228 [+] on Build GRCh38
          Mature miRNA Sequence: AUCCCUUGCAGGGGCUGUUGGGU
          Species: Human
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 001555
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000798, mir-623

          Mature miRNA Sequence:

          AUCCCUUGCAGGGGCUGUUGGGU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-623 MIMAT0003292

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-623
          Stem-loop Accession # MI0003637
          Stem-loop Sequence
          GUACACAGUAGAAGCAUCCCUUGCAGGGGCUGUUGGGUUGCAUCCUAAGCUGUGCUGGAGCUUCCCGAUGUACUCUGUAGAUGUCUUUGCACCUUCUG
          Chromosome Location Chr. 13 - 99356131 - 99356228 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003292
          miRBase ID: hsa-miR-623
          Mature miRNA Sequence: AUCCCUUGCAGGGGCUGUUGGGU
          Chromosome Location: Chr. 13 - 99356131 - 99356228 [+] on Build GRCh38


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Human Pool B v3.0


          Associated TaqMan® Array MicroRNA Card:

          TaqMan® Human MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panel:

          TaqMan® OpenArray® Human MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03304677_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11378
          Ambion® Pre-miR™ miRNA Precursor : PM11378
          mirVana® miRNA inhibitor : MH11378
          mirVana® miRNA mimic : MC11378
          TaqMan™ Advanced miRNA Assay : 479512_mir
          miRNA Card : TaqMan® Human MicroRNA B Cards v3.0
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          miRNA Pool : Megaplex™ RT Primers, Human Pool B v3.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® Array Human MicroRNA B Cards v3.0
          miRNA Card : TaqMan® Human MicroRNA B Cards v2.0
          FGMIR : TaqManTM Human MicroRNA Array B v3

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline