Search
Search

| Assay Name: | hsa-miR-483-3p |
|---|---|
| miRBase Accession Number: | MI0002467 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCACUCCUCUCCUCCCGUCUU |
| Species: | Human, Bovine |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000180, mir-483 |
Mature miRNA Sequence: | UCACUCCUCUCCUCCCGUCUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-483-3p | MIMAT0002173 |
| Bovine | bta-miR-483 | MIMAT0009327 |