Search
Search

| Assay Name: | hsa-miR-520c-3p |
|---|---|
| miRBase Accession Number: | MI0003158 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAAGUGCUUCCUUUUAGAGGGU |
| Species: | Human, Pongo pygmaeus |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | AAAGUGCUUCCUUUUAGAGGGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-520c-3p | MIMAT0002846 |
| Pongo pygmaeus | ppy-miR-520c-3p | MIMAT0015970 |