Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03306673_pri
          Assay Name:
          mmu-mir-106a
          Stem-loop ID:
          mmu-mir-106a mmu-mir-18b mmu-mir-20b
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.X: 52742503 - 52742567 [-] on Build GRCm38
          Stem-loop Location:
          Chr.X: 52742503 - 52742567 [-] on Build GRCm38 Chr.X: 52742331 - 52742413 [-] on Build GRCm38 Chr.X: 52742113 - 52742192 [-] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-106a-3p mmu-miR-106a-5p mmu-miR-18b-3p mmu-miR-18b-5p mmu-miR-20b-5p mmu-miR-20b-3p
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm03306673_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          72

          Chromosome Location:

          Chr. X - 52742706 - 52742706 [-] on 52742706 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-106a-3p MIMAT0017009
          Mouse mmu-miR-106a-5p MIMAT0000385
          Mouse mmu-miR-18b-3p MIMAT0017270
          Mouse mmu-miR-18b-5p MIMAT0004858
          Mouse mmu-miR-20b-5p MIMAT0003187
          Mouse mmu-miR-20b-3p MIMAT0004788

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-106a
          Stem-loop Accession # MI0000406
          Stem-loop Sequence
          AUGUCAAAGUGCUAACAGUGCAGGUAGCUUUUUGAGUUCUACUGCAGUGCCAGCACUUCUUACAU
          Chromosome Location Chr. X - 52742503 - 52742567 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017009
          miRBase ID: mmu-miR-106a-3p
          Mature miRNA Sequence: ACUGCAGUGCCAGCACUUCUUAC
          Chromosome Location: Chr. X - 52742503 - 52742567 [-] on Build GRCm38

          miRBase Accession #: MIMAT0000385
          miRBase ID: mmu-miR-106a-5p
          Mature miRNA Sequence: CAAAGUGCUAACAGUGCAGGUAG
          Chromosome Location: Chr. X - 52742503 - 52742567 [-] on Build GRCm38

          Gene Family ID MIPF0000001, mir-17
          Distance to Amplicon 102 bases
          Stem-loop ID mmu-mir-18b
          Stem-loop Accession # MI0005483
          Stem-loop Sequence
          UCUCUUGUGUUAAGGUGCAUCUAGUGCUGUUAGUGAAGCAGCUUACAAUCUACUGCCCUAAAUGCCCCUUCUCGCACAGGCUA
          Chromosome Location Chr. X - 52742331 - 52742413 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017270
          miRBase ID: mmu-miR-18b-3p
          Mature miRNA Sequence: UACUGCCCUAAAUGCCCCUUCU
          Chromosome Location: Chr. X - 52742331 - 52742413 [-] on Build GRCm38

          miRBase Accession #: MIMAT0004858
          miRBase ID: mmu-miR-18b-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCUGUUAG
          Chromosome Location: Chr. X - 52742331 - 52742413 [-] on Build GRCm38

          Gene Family ID MIPF0000001, mir-17
          Distance to Amplicon 256 bases
          Stem-loop ID mmu-mir-20b
          Stem-loop Accession # MI0003536
          Stem-loop Sequence
          CCUAGUAGUGCCAAAGUGCUCAUAGUGCAGGUAGUUUUUAUACCACUCUACUGCAGUGUGAGCACUUCUAGUACUCCUGG
          Chromosome Location Chr. X - 52742113 - 52742192 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003187
          miRBase ID: mmu-miR-20b-5p
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: Chr. X - 52742113 - 52742192 [-] on Build GRCm38

          miRBase Accession #: MIMAT0004788
          miRBase ID: mmu-miR-20b-3p
          Mature miRNA Sequence: ACUGCAGUGUGAGCACUUCUAG
          Chromosome Location: Chr. X - 52742113 - 52742192 [-] on Build GRCm38

          Gene Family ID MIPF0000001, mir-17
          Distance to Amplicon 477 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM20274
          Ambion® Pre-miR™ miRNA Precursor : PM20274
          TaqMan™ MicroRNA Assay : 462576_mat
          mirVana® miRNA inhibitor : MH20274
          mirVana® miRNA mimic : MC20274
          TaqMan™ Advanced miRNA Assay : mmu481514_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10975
          Ambion® Pre-miR™ miRNA Precursor : PM10975
          TaqMan™ MicroRNA Assay : 001014
          mirVana® miRNA inhibitor : MH10975
          mirVana® miRNA mimic : MC10975
          TaqMan™ Advanced miRNA Assay : rno477804_mir
          TaqMan™ Advanced miRNA Assay : 477804_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12726
          Ambion® Pre-miR™ miRNA Precursor : PM12726
          TaqMan™ MicroRNA Assay : 002459
          mirVana® miRNA inhibitor : MH12726
          mirVana® miRNA mimic : MC12726
          Ambion® Anti-miR™ miRNA Inhibitor : AM12633
          Ambion® Pre-miR™ miRNA Precursor : PM12633
          TaqMan™ MicroRNA Assay : 002524
          mirVana® miRNA inhibitor : MH12633
          mirVana® miRNA mimic : MC12633
          Ambion® Anti-miR™ miRNA Inhibitor : AM19657
          Ambion® Pre-miR™ miRNA Precursor : PM19657
          TaqMan™ MicroRNA Assay : 465227_mat
          mirVana® miRNA inhibitor : MH19657
          mirVana® miRNA mimic : MC19657
          TaqMan™ Advanced miRNA Assay : mmu481575_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12389
          Ambion® Pre-miR™ miRNA Precursor : PM12389
          TaqMan™ MicroRNA Assay : 002466
          mirVana® miRNA inhibitor : MH12389
          mirVana® miRNA mimic : MC12389


          miRBase Alias:

          Mouse: mmu-mir-18b (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b6cf8ccb7-qqcp6:80/100.66.76.64:80.
          git-commit: 76b2fce788d0185f5e22c122d85c407206860d42
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.0-2026-offline-prod