Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04351818_pri
          Assay Name:
          mmu-mir-7006
          Stem-loop ID:
          mmu-mir-7006
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.2: 181587900 - 181587968 [-] on Build GRCm38
          Stem-loop Location:
          Chr.2: 181587900 - 181587968 [-] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-7006-3p mmu-miR-7006-5p
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm04351818_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          65

          Chromosome Location:

          Chr. 2 - 181588126 - 181588126 [-] on 181588126 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-7006-3p MIMAT0027917
          Mouse mmu-miR-7006-5p MIMAT0027916

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-7006
          Stem-loop Accession # MI0022855
          Stem-loop Sequence
          CCAUGUGGGGGUGUUCAAGACUUGGGAACCUGGCUGAUUUCUUACCUGGUUUCUGACCUGGAUCCCCAG
          Chromosome Location Chr. 2 - 181587900 - 181587968 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0027917
          miRBase ID: mmu-miR-7006-3p
          Mature miRNA Sequence: UUUCUGACCUGGAUCCCCAG
          Chromosome Location: Chr. 2 - 181587900 - 181587968 [-] on Build GRCm38

          miRBase Accession #: MIMAT0027916
          miRBase ID: mmu-miR-7006-5p
          Mature miRNA Sequence: UGGGGGUGUUCAAGACUUGGGAACC
          Chromosome Location: Chr. 2 - 181587900 - 181587968 [-] on Build GRCm38

          Gene Family ID
          Distance to Amplicon 123 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM29377
          Ambion® Pre-miR™ miRNA Precursor : PM29377
          TaqMan™ MicroRNA Assay : 466593_mat
          mirVana® miRNA inhibitor : MH29377
          mirVana® miRNA mimic : MC29377
          TaqMan™ Advanced miRNA Assay : mmu482244_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM27769
          Ambion® Pre-miR™ miRNA Precursor : PM27769
          TaqMan™ MicroRNA Assay : 466308_mat
          mirVana® miRNA inhibitor : MH27769
          mirVana® miRNA mimic : MC27769
          TaqMan™ Advanced miRNA Assay : mmu482245_mir

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline