Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
ANKRD35
Entrez Gene ID
148741
Gene Name:
ankyrin repeat domain 35
Gene Aliases
-
Gene Chromosome Location
Chr.1: on Build GRCh38
UniGene
Hs.710624
Species:
Human
Primer Chromosome Location
Chr.1: 145872901-145872405 on Build GRCh38
View in UCSC browser
Amplicon Length
497
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00399911_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
ANKRD35
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_001280799.1
NP_001267728.1
-
-
-
NM_144698.4
NP_653299.4
6600695
c.2357T>C
p.L786P
4479460
c.2328C>T
p.S776S
1320034
c.1961G>A
p.R654Q
3801656
c.2142C>T
p.G714G
3801655
c.1875T>A
p.S625R
3418004
c.2115G>A
p.W705*
4661876
c.2289G>A
p.P763P
3984170
c.2072G>C
p.R691P
4661875
c.2128G>A
p.A710T
234201
c.2104C>T
p.R702*
1333600
c.2245C>T
p.R749W
2121040
c.1986G>A
p.A662A
4995258
c.2110C>A
p.L704M
5668902
c.2215C>A
p.R739R
5663943
c.2233C>A
p.Q745K
6206533
c.2323G>T
p.A775S
4021767
c.2341G>A
p.A781T
5641080
c.2143G>A
p.E715K
4021766
c.2095A>G
p.S699G
73676
c.1964A>T
p.E655V
4950547
c.1975delA
p.K659fs*18
4021765
c.1912C>T
p.R638W
4945187
c.2296G>C
p.E766Q
3862590
c.2175C>T
p.S725S
3473136
c.2287C>T
p.P763S
3473137
c.2288C>T
p.P763L
4591712
c.2278T>G
p.S760A
6861024
c.2147G>A
p.R716K
6365698
c.2135T>C
p.L712P
6363280
c.2258A>G
p.E753G
5146765
c.2215C>T
p.R739W
4535322
c.2179G>T
p.E727*
6360458
c.2206C>G
p.L736V
5716911
c.2116G>A
p.D706N
4142535
c.1974C>A
p.P658P
3704930
c.2145G>C
p.E715D
5518192
c.1985C>T
p.A662V
1238190
c.1940C>T
p.S647F
5881505
c.2046G>A
p.M682I
5843467
c.1930G>A
p.E644K
365966
c.1960C>T
p.R654W
895209
c.2272C>T
p.R758W
3473138
c.2342C>T
p.A781V
301679
c.2221C>A
p.R741R
895208
c.1946G>A
p.S649N
307660
c.1876A>T
p.N626Y
GenBank
AK057853.1
BAB71597.1
-
-
-
AK091120.1
BAC03587.1
-
-
-
AK294308.1
-
-
-
BC101618.1
AAI01619.1
-
-
-
BC101620.1
AAI01621.1
-
-
-
BQ636616.1
-
-
-
ENST00000355594.0
-
-
-
Phenotype
Literature Links:
ANKRD35
PubMed Links
SNP ID
Back To Top
More Information
Set Membership:
Between 400-700 bp
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0