Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
CIC
Entrez Gene ID
23152
Gene Name:
capicua transcriptional repressor
Gene Aliases
-
Gene Chromosome Location
Chr.19: on Build GRCh38
UniGene
Hs.388236
Species:
Human
Primer Chromosome Location
Chr.19: 42287520-42287665 on Build GRCh38
View in UCSC browser
Amplicon Length
146
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00517184_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
CIC
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_001304815.1
NP_001291744.1
-
-
-
NM_015125.4
NP_055940.3
-
-
-
XM_005258673.2
XP_005258730.1
5731279
c.692G>A
p.S231N
304189
c.699_701delCCT
p.I233_L234>M
235702
c.682C>T
p.R228W
235704
c.613A>C
p.N205H
132818
c.643C>T
p.R215W
7257526
c.694A>C
p.K232Q
303950
c.626T>G
p.I209S
5707042
c.652_656delGTCCA
p.V218fs*95
6905602
c.633C>A
p.S211R
132674
c.638G>C
p.R213P
5760555
c.679A>C
p.N227H
3971022
c.629T>G
p.F210C
6280160
c.666T>C
p.H222H
6959049
c.646G>T
p.A216S
3971024
c.703G>T
p.G235C
1737900
c.607C>G
p.P203A
235720
c.633C>G
p.S211R
6629183
c.661C>T
p.R221C
997306
c.662G>A
p.R221H
303949
c.604C>T
p.R202W
235712
c.605G>A
p.R202Q
6909987
c.676G>C
p.D226H
3971020
c.607C>T
p.P203S
5760550
c.630C>A
p.F210L
1737896
c.633_633delC
p.S211fs*46
4078713
c.603G>A
p.R201R
5731282
c.614A>T
p.N205I
5910996
c.668C>T
p.P223L
235724
c.611T>C
p.M204T
6938915
c.680A>T
p.N227I
6356171
c.687C>T
p.T229T
1716738
c.676G>T
p.D226Y
132673
c.601C>T
p.R201W
5726544
c.696G>A
p.K232K
132675
c.644G>A
p.R215Q
5047180
c.625A>T
p.I209F
3971023
c.701T>A
p.L234Q
5603195
c.686C>T
p.T229I
1737899
c.613_616AATG>T
p.N205_A206>S
997305
c.638G>A
p.R213Q
235721
c.682C>G
p.R228G
997307
c.683G>A
p.R228Q
5762251
c.602G>A
p.R201Q
6966041
c.595C>G
p.H199D
235709
c.619T>A
p.F207I
6629205
c.637C>T
p.R213W
243707
c.688G>A
p.V230I
1737895
c.630_632delCAG
p.S211del
235715
c.677A>G
p.D226G
6937450
c.621C>A
p.F207L
3971021
c.614A>G
p.N205S
XM_005258674.2
XP_005258731.1
-
-
-
XM_005258675.2
XP_005258732.1
-
-
-
XM_011526660.2
XP_011524962.1
-
-
-
XM_011526661.2
XP_011524963.1
-
-
-
XM_011526662.2
XP_011524964.1
-
-
-
XM_011526663.2
XP_011524965.1
-
-
-
XM_011526664.2
XP_011524966.1
-
-
-
XM_011526665.2
XP_011524967.1
-
-
-
XM_011526666.2
XP_011524968.1
-
-
-
XM_011526667.2
XP_011524969.1
-
-
-
GenBank
AB002304.1
BAA20765.1
-
-
-
AF363689.1
AAK73515.1
-
-
-
ENST00000160740.0
-
-
-
Phenotype
MIM: 612082
Literature Links:
CIC
PubMed Links
SNP ID
rs371965355
rs1040296890
rs748605354
rs778579143
rs1036745445
rs376814059
rs745501361
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.19: 42287598-42287598 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.19: 42287559-42287559 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.19: 42287556-42287556 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.19: 42287613-42287613 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.19: 42287544-42287544 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.19: 42287633-42287633 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.19: 42287623-42287623 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Back To Top
More Information
Set Membership:
Less than 200 bp
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
lung alveolus development
DNA binding
chromatin binding
protein binding
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0