Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ERG | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001136154.1 | NP_001129626.1 | - | - | - |
NM_001136155.1 | NP_001129627.1 | - | - | - | |
NM_001243428.1 | NP_001230357.1 | - | - | - | |
NM_001243429.1 | NP_001230358.1 | - | - | - | |
NM_001243432.2 | NP_001230361.1 | 4876213 | c.948C>T | p.G316G | |
4753311 | c.1045A>G | p.T349A | |||
2845108 | c.1005C>T | p.S335S | |||
1414063 | c.986C>T | p.S329L | |||
4681623 | c.1044G>C | p.M348I | |||
6371499 | c.976G>T | p.E326* | |||
6363198 | c.964C>T | p.Q322* | |||
3664559 | c.959T>A | p.L320H | |||
5392885 | c.1001C>T | p.S334F | |||
3550667 | c.969C>T | p.F323F | |||
4991420 | c.981C>T | p.L327L | |||
5906143 | c.1033G>A | p.E345K | |||
6976119 | c.1020A>T | p.E340D | |||
6973946 | c.958C>T | p.L320F | |||
NM_004449.4 | NP_004440.1 | 48372 | c.877C>A | p.Q293K | |
74541 | c.914C>T | p.S305L | |||
NM_182918.3 | NP_891548.1 | 4897463 | c.948C>T | p.F316F | |
6363199 | c.943C>T | p.Q315* | |||
4753312 | c.1024A>G | p.T342A | |||
6976120 | c.999A>T | p.E333D | |||
6973947 | c.937C>T | p.L313F | |||
5906144 | c.1012G>A | p.E338K | |||
4876214 | c.927C>T | p.G309G | |||
5392886 | c.980C>T | p.S327F | |||
4296151 | c.984C>T | p.S328S | |||
4947227 | c.965C>T | p.S322L | |||
4681624 | c.1023G>C | p.M341I | |||
4991421 | c.960C>T | p.L320L | |||
6371500 | c.955G>T | p.E319* | |||
4810512 | c.938T>A | p.L313H | |||
XM_011529486.2 | XP_011527788.1 | - | - | - | |
XM_017028287.1 | XP_016883776.1 | - | - | - | |
XM_017028288.1 | XP_016883777.1 | - | - | - | |
XM_017028289.1 | XP_016883778.1 | - | - | - | |
GenBank | AK297807.1 | - | - | - | |
AK300395.1 | - | - | - | ||
AK301277.1 | - | - | - | ||
AK303518.1 | - | - | - | ||
AK309469.1 | - | - | - | ||
AY204741.1 | AAP41719.1 | - | - | - | |
BC040168.1 | AAH40168.1 | - | - | - | |
ENST00000442448.0 | - | - | - | ||
KP100545.1 | - | - | - |
Literature Links: |
ERG PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |