Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TET2 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001127208.2 | NP_001120680.1 | 1716884 | c.1945C>T | p.Q649* |
211684 | c.1876C>T | p.Q626* | |||
4170162 | c.1955delA | p.Q652fs*48 | |||
110753 | c.1930C>T | p.Q644* | |||
144513 | c.1887_1887delC | p.H629fs*10 | |||
144516 | c.1842_1843insG | p.L615fs*23 | |||
340680 | c.1876C>G | p.Q626E | |||
5261949 | c.1959delC | p.Q654fs*46 | |||
1309588 | c.1882G>A | p.E628K | |||
1716773 | c.1912A>G | p.M638V | |||
6944574 | c.1942G>T | p.D648Y | |||
1644130 | c.1954_1954delC | p.Q652fs*48 | |||
5574229 | c.1918C>T | p.Q640* | |||
5945023 | c.1909G>T | p.E637* | |||
42084 | c.1924C>T | p.Q642* | |||
5990055 | c.1858_1859insA | p.Y620fs*1 | |||
211666 | c.1960C>T | p.Q654* | |||
46414 | c.1903C>T | p.Q635* | |||
219141 | c.1972_1972delC | p.H658fs*42 | |||
43521 | c.1843_1844insC | p.L615fs*23 | |||
5060046 | c.1849A>C | p.R617R | |||
7410084 | c.1968_1971delCTCA | p.S657fs*42 | |||
87093 | c.1860C>G | p.Y620* | |||
7409612 | c.1921delG | p.Q642fs*58 | |||
87091 | c.1847_1848insC | p.R617fs*21 | |||
5878964 | c.1937_1938insAGTGG | p.D648fs*54 | |||
6494932 | c.1880delT | p.L627fs*12 | |||
144540 | c.1909_1911GAA>AT | p.E637fs*2 | |||
4427469 | c.1925A>G | p.Q642R | |||
211683 | c.1860C>A | p.Y620* | |||
4170167 | c.1862delC | p.Q622fs*17 | |||
4996388 | c.1970C>G | p.S657* | |||
5944496 | c.1892_1893insA | p.Q632fs*6 | |||
4170035 | c.1888A>T | p.K630* | |||
110756 | c.1900_1901insTG | p.Y634fs*6 | |||
43478 | c.(1842_1844)insG | p.L615fs*23 | |||
7325154 | c.1970C>A | p.S657* | |||
41213 | c.1864C>T | p.Q622* | |||
5990104 | c.1958delT | p.F653fs*47 | |||
7409644 | c.1892C>A | p.S631* | |||
87092 | c.1851_1851delG | p.R617fs*22 | |||
6919876 | c.1882G>T | p.E628* | |||
5990077 | c.1906delG | p.V636fs*3 | |||
110758 | c.1894C>T | p.Q632* | |||
5878961 | c.1872delA | p.T625fs*14 | |||
6494934 | c.1884G>A | p.E628E | |||
42047 | c.1954C>T | p.Q652* | |||
4170048 | c.1843C>T | p.L615F | |||
3824892 | c.1966C>T | p.P656S | |||
NM_017628.4 | NP_060098.3 | 5990054 | c.1858_1859insA | p.Y620fs*1 | |
7325155 | c.1970C>A | p.S657* | |||
5574228 | c.1918C>T | p.Q640* | |||
6494933 | c.1884G>A | p.E628E | |||
4427468 | c.1925A>G | p.Q642R | |||
4383826 | c.1864C>T | p.Q622* | |||
1309587 | c.1882G>A | p.E628K | |||
5661635 | c.1876C>T | p.Q626* | |||
5878960 | c.1872delA | p.T625fs*14 | |||
5060045 | c.1849A>C | p.R617R | |||
4170055 | c.1960C>T | p.Q654* | |||
6919875 | c.1882G>T | p.E628* | |||
5878963 | c.1937_1938insAGTGG | p.D648fs*54 | |||
5990103 | c.1958delT | p.F653fs*47 | |||
3824891 | c.1966C>T | p.P656S | |||
4170047 | c.1843C>T | p.L615F | |||
6494931 | c.1880delT | p.L627fs*12 | |||
5990076 | c.1906delG | p.V636fs*3 | |||
4996387 | c.1970C>G | p.S657* | |||
4170034 | c.1888A>T | p.K630* | |||
4170028 | c.1954C>T | p.Q652* | |||
6944573 | c.1942G>T | p.D648Y | |||
5878962 | c.1903C>T | p.Q635* | |||
4170161 | c.1955delA | p.Q652fs*48 | |||
4170043 | c.1924C>T | p.Q642* | |||
1716883 | c.1945C>T | p.Q649* | |||
4170166 | c.1862delC | p.Q622fs*17 | |||
4385576 | c.1842_1843insG | p.L615fs*23 | |||
XM_005263082.2 | XP_005263139.1 | - | - | - | |
XM_006714242.3 | XP_006714305.1 | - | - | - | |
XM_017008319.1 | XP_016863808.1 | - | - | - | |
GenBank | AK000039.1 | BAA90898.1 | - | - | - |
AK027819.1 | BAB55391.1 | - | - | - | |
AK055149.1 | - | - | - | ||
BC150180.1 | - | - | - | ||
BC150201.1 | - | - | - | ||
BX640738.1 | CAE45851.1 | - | - | - | |
ENST00000380013.0 | - | - | - | ||
FM992369.1 | - | - | - |
TET2-AS1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NR_126420.1 | - | - | - | |
GenBank | AI401598.1 | - | - | - | |
BX100522.1 | - | - | - |
Literature Links: |
TET2 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |