Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TET2 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001127208.2 | NP_001120680.1 | 5708950 | c.296G>C | p.S99T |
3735040 | c.285_285delA | p.K95fs*18 | |||
6800073 | c.218G>A | p.R73H | |||
87069 | c.322C>T | p.Q108* | |||
6931474 | c.213_217delTAGTC | p.N71fs*5 | |||
1228965 | c.290C>T | p.T97I | |||
4383748 | c.220delG | p.V74fs*1 | |||
5944238 | c.209_213delAGAAT | p.N71fs*5 | |||
1049740 | c.268C>T | p.Q90* | |||
1228968 | c.260A>C | p.K87T | |||
4383752 | c.311delC | p.S104fs*9 | |||
6930061 | c.259delA | p.K87fs*8 | |||
1485573 | c.305_307delCTC | p.S102_L103>F | |||
43428 | c.238C>T | p.Q80* | |||
7409622 | c.328A>T | p.K110* | |||
732048 | c.223A>T | p.S75C | |||
6919736 | c.274G>C | p.G92R | |||
3916904 | c.274G>A | p.G92R | |||
6498606 | c.241_242insA | p.S82fs*2 | |||
211755 | c.318_331del14 | p.L107fs*8 | |||
6848515 | c.320T>A | p.L107H | |||
110763 | c.256_257insT | p.S86fs*14 | |||
1049741 | c.304_305delTC | p.S104fs*15 | |||
7184818 | c.282A>G | p.I94M | |||
6800075 | c.287_288insCACA | p.V98fs*3 | |||
5579709 | c.315G>A | p.G105G | |||
6924886 | c.179A>T | p.Y60F | |||
233565 | c.275G>A | p.G92E | |||
447196 | c.264T>C | p.C88C | |||
1018321 | c.291_294delAGTT | p.S99fs*13 | |||
4733145 | c.286C>T | p.R96C | |||
4383750 | c.307C>G | p.L103V | |||
6582096 | c.248G>A | p.R83K | |||
1049739 | c.241G>T | p.E81* | |||
4171286 | c.301C>A | p.P101T | |||
2270971 | c.323A>T | p.Q108L | |||
3916906 | c.313G>A | p.G105R | |||
5479692 | c.278G>A | p.G93E | |||
5878922 | c.155delC | p.K53fs*14 | |||
732047 | c.324G>A | p.Q108Q | |||
5400250 | c.257C>T | p.S86F | |||
NM_017628.4 | NP_060098.3 | 5479691 | c.278G>A | p.G93E | |
6800074 | c.287_288insCACA | p.V98fs*3 | |||
6582095 | c.248G>A | p.R83K | |||
1594217 | c.304_305delTC | p.S104fs*15 | |||
6930060 | c.259delA | p.K87fs*8 | |||
7184819 | c.282A>G | p.I94M | |||
6924885 | c.179A>T | p.Y60F | |||
5878921 | c.155delC | p.K53fs*14 | |||
6498605 | c.241_242insA | p.S82fs*2 | |||
4383749 | c.307C>G | p.L103V | |||
4733144 | c.286C>T | p.R96C | |||
1228967 | c.260A>C | p.K87T | |||
3916903 | c.274G>A | p.G92R | |||
1149829 | c.223A>T | p.S75C | |||
1485572 | c.305_307delCTC | p.S102_L103>F | |||
4171285 | c.301C>A | p.P101T | |||
1154266 | c.268C>T | p.Q90* | |||
6800072 | c.218G>A | p.R73H | |||
1594218 | c.241G>T | p.E81* | |||
5708949 | c.296G>C | p.S99T | |||
5400249 | c.257C>T | p.S86F | |||
4383747 | c.220delG | p.V74fs*1 | |||
6919735 | c.274G>C | p.G92R | |||
1228964 | c.290C>T | p.T97I | |||
4383751 | c.311delC | p.S104fs*9 | |||
6931473 | c.213_217delTAGTC | p.N71fs*5 | |||
1149830 | c.324G>A | p.Q108Q | |||
5579708 | c.315G>A | p.G105G | |||
3916905 | c.313G>A | p.G105R | |||
1485571 | c.264T>C | p.C88C | |||
XM_005263082.2 | XP_005263139.1 | - | - | - | |
XM_006714242.3 | XP_006714305.1 | - | - | - | |
XM_017008319.1 | XP_016863808.1 | - | - | - | |
GenBank | AI084057.1 | - | - | - | |
AK055149.1 | - | - | - | ||
BC150180.1 | - | - | - | ||
BC150201.1 | - | - | - | ||
BX640738.1 | CAE45851.1 | - | - | - | |
DA391215.1 | - | - | - | ||
DR005542.1 | - | - | - | ||
ENST00000380013.0 | - | - | - | ||
FM992369.1 | - | - | - |
TET2-AS1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NR_126420.1 | - | - | - | |
GenBank | AI401598.1 | - | - | - | |
BX100522.1 | - | - | - |
Literature Links: |
TET2 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |