Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
DNMT3A | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001320893.1 | NP_001307822.1 | - | - | - |
NM_022552.4 | NP_072046.2 | 4385519 | c.2422_2437del16 | p.T808fs*12 | |
5944897 | c.2450A>T | p.E817V | |||
7409533 | c.2478+9C>T | p.? | |||
5944195 | c.2418_2422delATCCA | p.S807fs*3 | |||
231548 | c.2409_2478del70 | p.R803fs*5 | |||
2911744 | c.2443C>A | p.L815M | |||
1318943 | c.2409G>C | p.R803S | |||
5878750 | c.2415G>C | p.L805F | |||
6927370 | c.2468G>A | p.R823K | |||
6091113 | c.2410C>T | p.P804S | |||
5708156 | c.2478G>T | p.K826N | |||
4766078 | c.2478+1delG | p.? | |||
4768018 | c.2458G>C | p.E820Q | |||
4963841 | c.2412G>T | p.P804P | |||
6907643 | c.2462A>G | p.H821R | |||
1583119 | c.2409_2422del14 | p.R803fs*4 | |||
4766074 | c.2409-1G>A | p.? | |||
99739 | c.2446C>T | p.Q816* | |||
6907614 | c.2443C>G | p.L815V | |||
1318945 | c.2478+2T>G | p.? | |||
6950203 | c.2409-1_2409GG>TT | p.? | |||
6906660 | c.2465G>A | p.G822D | |||
5500833 | c.2455C>G | p.L819V | |||
5944206 | c.2462delA | p.H821fs*4 | |||
6966097 | c.2411C>T | p.P804L | |||
4766079 | c.2478+1G>A | p.? | |||
7409803 | c.2451delG | p.E817fs*8 | |||
6907713 | c.2459A>G | p.E820G | |||
6907763 | c.2409-2A>G | p.? | |||
1583120 | c.2456T>C | p.L819P | |||
1583124 | c.2477A>G | p.K826R | |||
4991713 | c.2420C>T | p.S807F | |||
NM_153759.3 | NP_715640.2 | 6907615 | c.1876C>G | p.L626V | |
6907566 | c.1911+1G>A | p.? | |||
6927371 | c.1901G>A | p.R634K | |||
1318944 | c.1842G>C | p.R614S | |||
5878751 | c.1848G>C | p.L616F | |||
6907714 | c.1892A>G | p.E631G | |||
6091114 | c.1843C>T | p.P615S | |||
5708157 | c.1911G>T | p.K637N | |||
6966098 | c.1844C>T | p.P615L | |||
6907644 | c.1895A>G | p.H632R | |||
6950204 | c.1842-1_1842GG>TT | p.? | |||
5500834 | c.1888C>G | p.L630V | |||
4768019 | c.1891G>C | p.E631Q | |||
6907764 | c.1842-2A>G | p.? | |||
4991714 | c.1853C>T | p.S618F | |||
4963842 | c.1845G>T | p.P615P | |||
1318946 | c.1911+2T>G | p.? | |||
2911745 | c.1876C>A | p.L626M | |||
4385520 | c.1855_1870del16 | p.T619fs*12 | |||
NM_175629.2 | NP_783328.1 | - | - | - | |
NR_135490.1 | - | - | - | ||
XM_005264175.4 | XP_005264232.1 | - | - | - | |
XM_005264177.4 | XP_005264234.1 | - | - | - | |
XM_011532662.2 | XP_011530964.1 | - | - | - | |
XM_011532663.2 | XP_011530965.1 | - | - | - | |
XM_011532666.2 | XP_011530968.1 | - | - | - | |
XM_011532667.2 | XP_011530969.1 | - | - | - | |
XM_017003526.1 | XP_016859015.1 | - | - | - | |
XM_017003527.1 | XP_016859016.1 | - | - | - | |
GenBank | AB208833.1 | BAD92070.1 | - | - | - |
AF067972.2 | AAD33084.2 | - | - | - | |
AF331856.1 | AAL57039.1 | - | - | - | |
AF480163.1 | AAN40037.1 | - | - | - | |
AK025230.1 | - | - | - | ||
BC018214.1 | AAH18214.1 | - | - | - | |
BC023612.2 | AAH23612.1 | - | - | - | |
BC043617.1 | AAH43617.1 | - | - | - | |
ENST00000321117.0 | - | - | - |
Literature Links: |
DNMT3A PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
Set Membership: |
Less than 200 bp |