Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TET2 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001127208.2 | NP_001120680.1 | 5708954 | c.1051G>T | p.E351* |
41878 | c.1147C>T | p.Q383* | |||
231578 | c.1037T>C | p.L346P | |||
5523998 | c.1172C>T | p.S391F | |||
5878935 | c.1107_1108delGT | p.Y370fs*5 | |||
211678 | c.1110T>A | p.Y370* | |||
100060 | c.1033_1036delAAGC | p.K345fs*1 | |||
5878939 | c.1123G>T | p.E375* | |||
96928 | c.(1176_1179)insT | p.(S393_A394)fs*(49_50) | |||
4774908 | c.1091G>A | p.G364D | |||
6968905 | c.1124A>G | p.E375G | |||
4121121 | c.1150A>G | p.S384G | |||
5865527 | c.1067C>T | p.S356F | |||
1426205 | c.1040C>T | p.A347V | |||
2952706 | c.1041G>A | p.A347A | |||
3916908 | c.1173C>T | p.S391S | |||
120173 | c.1090_1091GG>T | p.G364fs*8 | |||
7256437 | c.1095C>A | p.G365G | |||
1169697 | c.1061C>A | p.S354* | |||
5945711 | c.1062_1062delA | p.G355fs*17 | |||
4383778 | c.1059T>A | p.C353* | |||
4383782 | c.1103delA | p.E368fs*4 | |||
5020142 | c.1088C>T | p.P363L | |||
87083 | c.1137_1138TT>C | p.Y380fs*47 | |||
87081 | c.1037_1038insAGCT | p.G349fs*3 | |||
43440 | c.1061C>G | p.S354* | |||
6907524 | c.1024G>T | p.G342* | |||
6977794 | c.1102G>C | p.E368Q | |||
1228970 | c.1056C>A | p.F352L | |||
5878941 | c.1174delT | p.S393fs*34 | |||
7409834 | c.1054delT | p.F352fs*20 | |||
5577117 | c.1167G>A | p.K389K | |||
5944492 | c.1171_1172insT | p.S391fs*52 | |||
1737965 | c.1072A>G | p.S358G | |||
6494926 | c.1064G>A | p.G355D | |||
5989620 | c.1024delG | p.G342fs*5 | |||
4766102 | c.1169_1169delA | p.D390fs*37 | |||
211679 | c.1157_1157delT | p.V386fs*41 | |||
6022443 | c.1169_1170ins14 | p.D390fs*42 | |||
5878937 | c.1113delA | p.K372fs*5 | |||
7410060 | c.1138_1156del19 | p.Y380fs*41 | |||
4121123 | c.1177T>G | p.S393A | |||
6987367 | c.1142_1143insA | p.F381fs*9 | |||
6942646 | c.1105C>T | p.R369W | |||
5641942 | c.1161C>T | p.F387F | |||
43477 | c.(1060_1062)insA | p.(S354_G355)fs*? | |||
7409614 | c.1058delG | p.C353fs*19 | |||
4383780 | c.1070delG | p.S357fs*15 | |||
7413175 | c.1177_1177delT | p.S393fs*34 | |||
4383784 | c.1164delT | p.K389fs*38 | |||
87082 | c.1118_1122delAAAAT | p.Q373fs*15 | |||
6968041 | c.1031C>A | p.T344K | |||
NM_017628.4 | NP_060098.3 | 4383777 | c.1059T>A | p.C353* | |
6494925 | c.1064G>A | p.G355D | |||
5878936 | c.1113delA | p.K372fs*5 | |||
4383781 | c.1103delA | p.E368fs*4 | |||
6977793 | c.1102G>C | p.E368Q | |||
5865526 | c.1067C>T | p.S356F | |||
5989619 | c.1024delG | p.G342fs*5 | |||
2952705 | c.1041G>A | p.A347A | |||
6968040 | c.1031C>A | p.T344K | |||
6942645 | c.1105C>T | p.R369W | |||
5641941 | c.1161C>T | p.F387F | |||
6987366 | c.1142_1143insA | p.F381fs*9 | |||
6907523 | c.1024G>T | p.G342* | |||
4121120 | c.1150A>G | p.S384G | |||
5020141 | c.1088C>T | p.P363L | |||
1426204 | c.1040C>T | p.A347V | |||
5878940 | c.1174delT | p.S393fs*34 | |||
4170051 | c.1061C>G | p.S354* | |||
3916907 | c.1173C>T | p.S391S | |||
5459079 | c.1072A>G | p.S358G | |||
4383779 | c.1070delG | p.S357fs*15 | |||
5523997 | c.1172C>T | p.S391F | |||
5878934 | c.1107_1108delGT | p.Y370fs*5 | |||
5708953 | c.1051G>T | p.E351* | |||
4383783 | c.1164delT | p.K389fs*38 | |||
4774907 | c.1091G>A | p.G364D | |||
6968904 | c.1124A>G | p.E375G | |||
7256438 | c.1095C>A | p.G365G | |||
1228969 | c.1056C>A | p.F352L | |||
4121122 | c.1177T>G | p.S393A | |||
5878938 | c.1123G>T | p.E375* | |||
4170040 | c.1147C>T | p.Q383* | |||
5577116 | c.1167G>A | p.K389K | |||
XM_005263082.2 | XP_005263139.1 | - | - | - | |
XM_006714242.3 | XP_006714305.1 | - | - | - | |
XM_017008319.1 | XP_016863808.1 | - | - | - | |
GenBank | AK000039.1 | BAA90898.1 | - | - | - |
AK055149.1 | - | - | - | ||
BC150180.1 | - | - | - | ||
BC150201.1 | - | - | - | ||
BX640738.1 | CAE45851.1 | - | - | - | |
ENST00000380013.0 | - | - | - | ||
FM992369.1 | - | - | - |
TET2-AS1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NR_126420.1 | - | - | - | |
GenBank | AI401598.1 | - | - | - | |
BX100522.1 | - | - | - |
Literature Links: |
TET2 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |