Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FGFR1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001174063.1 | NP_001167534.1 | - | - | - |
NM_001174064.1 | NP_001167535.1 | 3900040 | c.762C>T | p.P254P | |
1137869 | c.763G>T | p.A255S | |||
3648898 | c.801C>T | p.F267F | |||
2960509 | c.778G>T | p.A260S | |||
3785990 | c.725G>A | p.R242Q | |||
1292691 | c.792C>T | p.N264N | |||
5420180 | c.822C>T | p.D274D | |||
7303572 | c.722-8T>G | p.? | |||
3900034 | c.818G>A | p.S273N | |||
3900046 | c.724C>T | p.R242W | |||
3648904 | c.729C>T | p.S243S | |||
2960524 | c.728C>T | p.S243F | |||
3395325 | c.736C>T | p.R246W | |||
3413002 | c.737G>T | p.R246L | |||
6663594 | c.760C>T | p.P254S | |||
NM_001174065.1 | NP_001167536.1 | - | - | - | |
NM_001174066.1 | NP_001167537.1 | - | - | - | |
NM_001174067.1 | NP_001167538.1 | 5420181 | c.579C>T | p.D193D | |
2960525 | c.485C>T | p.S162F | |||
334851 | c.495G>T | p.R165R | |||
3413003 | c.494G>T | p.R165L | |||
7303573 | c.479-8T>G | p.? | |||
3648905 | c.486C>T | p.S162S | |||
3785991 | c.482G>A | p.R161Q | |||
3900041 | c.519C>T | p.P173P | |||
1292692 | c.549C>T | p.N183N | |||
486443 | c.520G>T | p.A174S | |||
2960510 | c.535G>T | p.A179S | |||
3395326 | c.493C>T | p.R165W | |||
3648899 | c.558C>T | p.F186F | |||
6663595 | c.517C>T | p.P173S | |||
3900035 | c.575G>A | p.S192N | |||
3900047 | c.481C>T | p.R161W | |||
NM_015850.3 | NP_056934.2 | - | - | - | |
NM_023105.2 | NP_075593.1 | - | - | - | |
NM_023106.2 | NP_075594.1 | - | - | - | |
NM_023110.2 | NP_075598.2 | 7347043 | c.830G>A | p.C277Y | |
12834 | c.754C>A | p.P252T | |||
3785989 | c.749G>A | p.R250Q | |||
6663596 | c.784C>T | p.P262S | |||
486442 | c.787G>T | p.A263S | |||
3900037 | c.842G>A | p.S281N | |||
2960511 | c.802G>T | p.A268S | |||
3900049 | c.748C>T | p.R250W | |||
2960526 | c.752C>T | p.S251F | |||
3648906 | c.753C>T | p.S251S | |||
3395324 | c.760C>T | p.R254W | |||
3413001 | c.761G>T | p.R254L | |||
334852 | c.762G>T | p.R254R | |||
3900033 | c.842G>A | p.S281N | |||
6842042 | c.757C>T | p.H253Y | |||
6196601 | c.808G>T | p.G270C | |||
5420179 | c.846C>T | p.D282D | |||
7303574 | c.746-8T>G | p.? | |||
3648897 | c.825C>T | p.F275F | |||
3900045 | c.748C>T | p.R250W | |||
2960508 | c.802G>T | p.A268S | |||
1292693 | c.816C>T | p.N272N | |||
1292690 | c.816C>T | p.N272N | |||
334850 | c.762G>T | p.R254R | |||
3648900 | c.825C>T | p.F275F | |||
5420182 | c.846C>T | p.D282D | |||
3900043 | c.786C>T | p.P262P | |||
2960521 | c.754C>T | p.P252S | |||
3785992 | c.749G>A | p.R250Q | |||
3900039 | c.786C>T | p.P262P | |||
3395327 | c.760C>T | p.R254W | |||
3413004 | c.761G>T | p.R254L | |||
6663593 | c.784C>T | p.P262S | |||
7303571 | c.746-8T>G | p.? | |||
486444 | c.787G>T | p.A263S | |||
2960523 | c.752C>T | p.S251F | |||
3648903 | c.753C>T | p.S251S | |||
XM_006716303.2 | XP_006716366.1 | - | - | - | |
XM_006716304.1 | XP_006716367.1 | - | - | - | |
XM_006716306.2 | XP_006716369.1 | - | - | - | |
XM_006716307.1 | XP_006716370.1 | - | - | - | |
XM_006716309.3 | XP_006716372.1 | - | - | - | |
XM_006716310.2 | XP_006716373.1 | - | - | - | |
XM_006716311.1 | XP_006716374.1 | - | - | - | |
XM_006716312.1 | XP_006716375.1 | - | - | - | |
XM_006716313.2 | XP_006716376.1 | - | - | - | |
XM_006716314.1 | XP_006716377.1 | - | - | - | |
XM_011544443.2 | XP_011542745.1 | - | - | - | |
XM_011544444.1 | XP_011542746.1 | - | - | - | |
XM_011544445.2 | XP_011542747.1 | - | - | - | |
XM_011544446.2 | XP_011542748.1 | - | - | - | |
XM_011544447.2 | XP_011542749.1 | - | - | - | |
XM_011544448.1 | XP_011542750.1 | - | - | - | |
XM_011544449.1 | XP_011542751.1 | - | - | - | |
XM_011544450.2 | XP_011542752.1 | - | - | - | |
XM_011544451.1 | XP_011542753.1 | - | - | - | |
XM_011544452.2 | XP_011542754.1 | - | - | - | |
XM_017013219.1 | XP_016868708.1 | - | - | - | |
XM_017013220.1 | XP_016868709.1 | - | - | - | |
XM_017013221.1 | XP_016868710.1 | - | - | - | |
XM_017013222.1 | XP_016868711.1 | - | - | - | |
XM_017013223.1 | XP_016868712.1 | - | - | - | |
XM_017013224.1 | XP_016868713.1 | - | - | - | |
XM_017013225.1 | XP_016868714.1 | - | - | - | |
XM_017013226.1 | XP_016868715.1 | - | - | - | |
XM_017013227.1 | XP_016868716.1 | - | - | - | |
XM_017013228.1 | XP_016868717.1 | - | - | - | |
XM_017013229.1 | XP_016868718.1 | - | - | - | |
XM_017013230.1 | XP_016868719.1 | - | - | - | |
XM_017013231.1 | XP_016868720.1 | - | - | - | |
GenBank | AB208919.1 | BAD92156.1 | - | - | - |
AK222718.1 | BAD96438.1 | - | - | - | |
AK223126.1 | BAD96846.1 | - | - | - | |
AK291754.1 | - | - | - | ||
AK292470.1 | - | - | - | ||
AK309947.1 | - | - | - | ||
BC015035.1 | AAH15035.1 | - | - | - | |
BC018128.1 | AAH18128.1 | - | - | - | |
BC091494.1 | AAH91494.1 | - | - | - | |
BX649167.1 | - | - | - | ||
ENST00000447712.0 | - | - | - | ||
EU826596.1 | - | - | - | ||
EU826597.1 | - | - | - | ||
FJ809916.1 | - | - | - | ||
FJ809917.1 | - | - | - | ||
KU178066.1 | - | - | - | ||
KU178067.1 | - | - | - | ||
KU178068.1 | - | - | - | ||
KU178069.1 | - | - | - | ||
KU178070.1 | - | - | - | ||
M34185.1 | AAA35836.1 | - | - | - | |
M34186.1 | AAA35837.1 | - | - | - | |
M34187.1 | AAA35838.1 | - | - | - | |
M34188.1 | AAA35839.1 | - | - | - | |
M34641.1 | AAA35835.1 | - | - | - | |
M37722.1 | AAA75007.1 | - | - | - | |
M60485.1 | AAA35840.1 | - | - | - | |
M63887.1 | AAA35958.1 | - | - | - | |
M63888.1 | AAA35959.1 | - | - | - | |
M63889.1 | AAA35960.1 | - | - | - | |
S76658.1 | AAD14230.1 | - | - | - | |
X51803.1 | CAA36101.1 | - | - | - | |
X52833.1 | CAA37015.1 | - | - | - | |
X57119.1 | CAA40401.1 | - | - | - | |
X57120.1 | CAA40402.1 | - | - | - | |
X57121.1 | CAA40403.1 | - | - | - | |
X57122.1 | CAA40404.1 | - | - | - | |
X66945.1 | CAA47375.1 | - | - | - | |
Y00665.1 | CAA68679.1 | - | - | - |
Literature Links: |
FGFR1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |