Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ESRRG | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001134285.2 | NP_001127757.1 | - | - | - |
NM_001243505.1 | NP_001230434.1 | - | - | - | |
NM_001243506.1 | NP_001230435.1 | - | - | - | |
NM_001243507.1 | NP_001230436.1 | - | - | - | |
NM_001243509.1 | NP_001230438.1 | - | - | - | |
NM_001243510.2 | NP_001230439.1 | - | - | - | |
NM_001243511.2 | NP_001230440.1 | - | - | - | |
NM_001243512.1 | NP_001230441.1 | - | - | - | |
NM_001243513.1 | NP_001230442.1 | - | - | - | |
NM_001243514.1 | NP_001230443.1 | - | - | - | |
NM_001243515.1 | NP_001230444.1 | 7234194 | c.653A>G | p.D218G | |
3400331 | c.652G>C | p.D218H | |||
3376973 | c.689C>T | p.P230L | |||
4231948 | c.690G>A | p.P230P | |||
4231945 | c.704C>T | p.A235V | |||
1958536 | c.633G>A | p.L211L | |||
4860078 | c.682G>C | p.A228P | |||
NM_001243518.1 | NP_001230447.1 | - | - | - | |
NM_001243519.1 | NP_001230448.1 | - | - | - | |
NM_001438.3 | NP_001429.2 | 678524 | c.730G>C | p.A244P | |
7234193 | c.701A>G | p.Y234C | |||
1958530 | c.752C>T | p.A251V | |||
280872 | c.721T>A | p.L241M | |||
3376972 | c.737C>T | p.P246L | |||
532581 | c.738G>A | p.P246P | |||
2156182 | c.701-1G>C | p.? | |||
NM_206594.2 | NP_996317.1 | - | - | - | |
NM_206595.2 | NP_996318.1 | - | - | - | |
XM_011509265.2 | XP_011507567.1 | - | - | - | |
XM_011509266.2 | XP_011507568.1 | - | - | - | |
XM_011509267.1 | XP_011507569.1 | - | - | - | |
XM_011509268.2 | XP_011507570.1 | - | - | - | |
XM_011509269.2 | XP_011507571.1 | - | - | - | |
XM_011509270.1 | XP_011507572.1 | - | - | - | |
XM_011509271.2 | XP_011507573.1 | - | - | - | |
XM_011509274.1 | XP_011507576.1 | - | - | - | |
XM_011509275.1 | XP_011507577.1 | - | - | - | |
XM_011509276.1 | XP_011507578.1 | - | - | - | |
XM_011509277.1 | XP_011507579.1 | - | - | - | |
XM_011509278.1 | XP_011507580.1 | - | - | - | |
XM_011509279.1 | XP_011507581.1 | - | - | - | |
XM_011509280.2 | XP_011507582.1 | - | - | - | |
XM_017000621.1 | XP_016856110.1 | - | - | - | |
XM_017000622.1 | XP_016856111.1 | - | - | - | |
XM_017000623.1 | XP_016856112.1 | - | - | - | |
XM_017000624.1 | XP_016856113.1 | - | - | - | |
XM_017000625.1 | XP_016856114.1 | - | - | - | |
XM_017000626.1 | XP_016856115.1 | - | - | - | |
XM_017000627.1 | XP_016856116.1 | - | - | - | |
XM_017000628.1 | XP_016856117.1 | - | - | - | |
XM_017000629.1 | XP_016856118.1 | - | - | - | |
XM_017000630.1 | XP_016856119.1 | - | - | - | |
XM_017000631.1 | XP_016856120.1 | - | - | - | |
XM_017000632.1 | XP_016856121.1 | - | - | - | |
XM_017000633.1 | XP_016856122.1 | - | - | - | |
XM_017000634.1 | XP_016856123.1 | - | - | - | |
XM_017000635.1 | XP_016856124.1 | - | - | - | |
XM_017000636.1 | XP_016856125.1 | - | - | - | |
XM_017000637.1 | XP_016856126.1 | - | - | - | |
XM_017000638.1 | XP_016856127.1 | - | - | - | |
XM_017000639.1 | XP_016856128.1 | - | - | - | |
XM_017000640.1 | XP_016856129.1 | - | - | - | |
XM_017000641.1 | XP_016856130.1 | - | - | - | |
XM_017000642.1 | XP_016856131.1 | - | - | - | |
XM_017000643.1 | XP_016856132.1 | - | - | - | |
XM_017000644.1 | XP_016856133.1 | - | - | - | |
XM_017000645.1 | XP_016856134.1 | - | - | - | |
XM_017000646.1 | XP_016856135.1 | - | - | - | |
XM_017000647.1 | XP_016856136.1 | - | - | - | |
XM_017000648.1 | XP_016856137.1 | - | - | - | |
XM_017000649.1 | XP_016856138.1 | - | - | - | |
GenBank | AB020639.1 | - | - | - | |
AB307715.1 | - | - | - | ||
AB362218.1 | - | - | - | ||
AF058291.1 | AAC39899.1 | - | - | - | |
AF094518.1 | AAC99410.1 | - | - | - | |
AK131193.1 | - | - | - | ||
AK290945.1 | - | - | - | ||
AK291028.1 | - | - | - | ||
AK291647.1 | - | - | - | ||
AK298851.1 | - | - | - | ||
AY388456.1 | AAQ93376.1 | - | - | - | |
AY388457.1 | AAQ93377.1 | - | - | - | |
AY388458.1 | AAQ93378.1 | - | - | - | |
AY388459.1 | AAQ93379.1 | - | - | - | |
AY388460.1 | AAQ93380.1 | - | - | - | |
AY388461.1 | AAQ93381.1 | - | - | - | |
BC008218.2 | AAH08218.1 | - | - | - | |
BC064700.1 | AAH64700.1 | - | - | - | |
BM723962.1 | - | - | - | ||
BX647076.1 | - | - | - | ||
CR749497.1 | CAH18320.1 | - | - | - | |
ENST00000408911.0 | - | - | - | ||
HQ692853.1 | - | - | - | ||
HQ692854.1 | - | - | - |
Literature Links: |
ESRRG PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
Set Membership: |
Between 200-400 bp |