Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
ANKRD34A
POLR3GL
Entrez Gene ID
284615
84265
Gene Name:
ankyrin repeat domain 34A
RNA polymerase III subunit G like
Gene Aliases
ANKRD34
RPC32HOM, flj32422
Gene Chromosome Location
Chr.1: on Build GRCh38
Chr.1: on Build GRCh38
UniGene
Hs.620591
Hs.591456
Species:
Human
Primer Chromosome Location
Chr.1: 145961703-145961430 on Build GRCh38
View in UCSC browser
Amplicon Length
274
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00607069_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
ANKRD34A
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_001039888.3
NP_001034977.1
-
-
-
GenBank
AK094282.1
-
-
-
AK128050.1
-
-
-
DA081864.1
-
-
-
ENST00000323397.0
1747699
c.284C>G
p.S95W
1667888
c.203C>T
p.A68V
5081855
c.255C>T
p.C85C
4021743
c.65G>A
p.R22H
6600673
c.108G>A
p.A36A
4021744
c.301G>A
p.G101S
5881503
c.289C>G
p.L97V
674818
c.274G>T
p.A92S
1182918
c.236C>T
p.T79M
5735604
c.145C>T
p.R49C
5704903
c.118A>G
p.T40A
2120825
c.224G>A
p.R75Q
423688
c.327C>T
p.H109H
4762346
c.302G>A
p.G101D
4661864
c.322G>T
p.D108Y
1293160
c.193G>A
p.E65K
4661862
c.122C>T
p.A41V
7110411
c.243C>A
p.L81L
1294930
c.284C>T
p.S95L
4661863
c.297C>A
p.A99A
3399667
c.233G>A
p.R78H
4661861
c.114G>A
p.G38G
6540805
c.319C>T
p.R107*
4888520
c.268G>A
p.G90S
6357056
c.270C>T
p.G90G
1333564
c.111delG
p.E39fs*4
1333565
c.271G>A
p.A91T
6445285
c.210C>A
p.P70P
1333563
c.76G>A
p.E26K
Phenotype
POLR3GL
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
XM_005277431.4
XP_005277488.1
-
-
-
Phenotype
Literature Links:
ANKRD34A
PubMed Links
SNP ID
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
DNA-directed RNA polymerase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase III promoter
transcription initiation from RNA polymerase III promoter
positive regulation of type I interferon production
RNA polymerase III activity
protein binding
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0