Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ESR1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_000125.3 | NP_000116.2 | - | - | - |
NM_001122740.1 | NP_001116212.1 | - | - | - | |
NM_001122741.1 | NP_001116213.1 | - | - | - | |
NM_001122742.1 | NP_001116214.1 | 3019458 | c.542A>C | p.E181A | |
4992439 | c.630G>A | p.K210K | |||
6906869 | c.640C>G | p.Q214E | |||
1205680 | c.632G>T | p.R211I | |||
740580 | c.512A>T | p.K171M | |||
740577 | c.587A>G | p.H196R | |||
6957990 | c.619G>A | p.A207T | |||
2156042 | c.496G>T | p.A166S | |||
6930803 | c.547C>G | p.R183G | |||
6954697 | c.608A>T | p.E203V | |||
4845847 | c.617A>G | p.K206R | |||
6995431 | c.568G>C | p.D190H | |||
4503501 | c.640C>T | p.Q214* | |||
1568008 | c.568G>A | p.D190N | |||
5975822 | c.508G>T | p.D170Y | |||
1074617 | c.571T>C | p.Y191H | |||
4428864 | c.597C>T | p.V199V | |||
333038 | c.514G>T | p.G172* | |||
6656385 | c.568G>T | p.D190Y | |||
3019459 | c.544A>G | p.T182A | |||
5659269 | c.529G>T | p.E177* | |||
3019455 | c.473G>A | p.R158H | |||
6909339 | c.476A>T | p.Q159L | |||
740578 | c.513G>A | p.K171K | |||
1205681 | c.643G>T | p.G215* | |||
4992437 | c.624C>T | p.F208F | |||
206400 | c.469C>T | p.R157* | |||
6547751 | c.522G>T | p.M174I | |||
1599950 | c.472C>T | p.R158C | |||
6919266 | c.470G>A | p.R157Q | |||
6441565 | c.514G>A | p.G172R | |||
3430122 | c.453-1G>T | p.? | |||
7344069 | c.467A>C | p.N156T | |||
6916263 | c.617A>C | p.K206T | |||
6981202 | c.547C>T | p.R183C | |||
6944548 | c.633A>T | p.R211S | |||
6382421 | c.614G>A | p.C205Y | |||
6980799 | c.479G>A | p.G160D | |||
NM_001291230.1 | NP_001278159.1 | - | - | - | |
NM_001291241.1 | NP_001278170.1 | - | - | - | |
NM_001328100.1 | NP_001315029.1 | - | - | - | |
XM_006715374.3 | XP_006715437.1 | - | - | - | |
XM_006715375.3 | XP_006715438.1 | - | - | - | |
XM_011535543.2 | XP_011533845.1 | - | - | - | |
XM_011535544.2 | XP_011533846.1 | - | - | - | |
XM_011535545.2 | XP_011533847.1 | - | - | - | |
XM_011535547.2 | XP_011533849.1 | - | - | - | |
XM_017010376.1 | XP_016865865.1 | - | - | - | |
XM_017010377.1 | XP_016865866.1 | - | - | - | |
XM_017010378.1 | XP_016865867.1 | - | - | - | |
XM_017010379.1 | XP_016865868.1 | - | - | - | |
XM_017010380.1 | XP_016865869.1 | - | - | - | |
XM_017010381.1 | XP_016865870.1 | - | - | - | |
XM_017010382.1 | XP_016865871.1 | - | - | - | |
GenBank | AB307713.1 | - | - | - | |
AB588629.1 | - | - | - | ||
AB588630.1 | - | - | - | ||
AB685814.1 | - | - | - | ||
AB685815.1 | - | - | - | ||
AB685817.1 | - | - | - | ||
AB685818.1 | - | - | - | ||
AB685819.1 | - | - | - | ||
AB685820.1 | - | - | - | ||
AF258449.1 | AAG41358.1 | - | - | - | |
AF258450.1 | AAG41359.1 | - | - | - | |
AF258451.1 | AAG41360.1 | - | - | - | |
AK293019.1 | - | - | - | ||
AK304833.1 | - | - | - | ||
AK309580.1 | - | - | - | ||
AK313644.1 | - | - | - | ||
BC128573.1 | - | - | - | ||
BC128574.1 | - | - | - | ||
BX640939.1 | CAE45969.1 | - | - | - | |
EF491789.1 | - | - | - | ||
ENST00000440973.0 | - | - | - | ||
EU921455.1 | - | - | - | ||
EU930361.1 | - | - | - | ||
EU982838.1 | - | - | - | ||
EU982839.1 | - | - | - | ||
EU982840.1 | - | - | - | ||
FJ011556.1 | - | - | - | ||
FJ011557.1 | - | - | - | ||
FJ011559.1 | - | - | - | ||
FJ011560.1 | - | - | - | ||
FJ147552.1 | - | - | - | ||
FJ147553.1 | - | - | - | ||
FJ147554.1 | - | - | - | ||
FJ147555.1 | - | - | - | ||
FJ147556.1 | - | - | - | ||
FJ147557.1 | - | - | - | ||
FJ147558.1 | - | - | - | ||
FJ169500.1 | - | - | - | ||
FJ169501.1 | - | - | - | ||
FJ169503.1 | - | - | - | ||
FJ169504.1 | - | - | - | ||
FJ668387.1 | - | - | - | ||
FJ712305.1 | - | - | - | ||
FJ712306.1 | - | - | - | ||
FJ712307.1 | - | - | - | ||
FJ716812.1 | - | - | - | ||
FJ716813.1 | - | - | - | ||
FJ716815.1 | - | - | - | ||
GQ366704.1 | - | - | - | ||
GQ389792.1 | - | - | - | ||
GQ411250.1 | - | - | - | ||
GQ411251.1 | - | - | - | ||
GQ411252.1 | - | - | - | ||
GQ411254.1 | - | - | - | ||
GQ411256.1 | - | - | - | ||
GQ411258.1 | - | - | - | ||
GQ438789.1 | - | - | - | ||
GQ438790.1 | - | - | - | ||
GQ438793.1 | - | - | - | ||
JF810888.1 | - | - | - | ||
LC120323.1 | - | - | - | ||
LC120324.1 | - | - | - | ||
LC120325.1 | - | - | - | ||
LC120326.1 | - | - | - | ||
LC120327.1 | - | - | - | ||
LC120328.1 | - | - | - | ||
LC120329.1 | - | - | - | ||
LC120330.1 | - | - | - | ||
M12674.1 | AAA52399.1 | - | - | - | |
M69297.1 | AAA58462.1 | - | - | - | |
X03635.1 | CAA27284.1 | - | - | - | |
X86816.1 | - | - | - |
Literature Links: |
ESR1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
Set Membership: |
Between 200-400 bp |