Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
EPB41 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001166005.1 | NP_001159477.1 | 6390562 | c.958T>C | p.C320R |
426041 | c.1070C>G | p.P357R | |||
6364391 | c.925C>A | p.L309I | |||
6363262 | c.1039G>T | p.V347L | |||
5214502 | c.1056delT | p.F353fs*25 | |||
6654585 | c.1054G>T | p.D352Y | |||
908012 | c.1027G>T | p.E343* | |||
4977760 | c.1002G>T | p.Q334H | |||
7428412 | c.1071G>A | p.P357P | |||
1251240 | c.999C>T | p.I333I | |||
2152925 | c.1021G>A | p.D341N | |||
6654557 | c.928C>T | p.R310W | |||
7308788 | c.906-4G>C | p.? | |||
5993152 | c.1038C>T | p.G346G | |||
4681158 | c.919C>T | p.L307F | |||
NM_001166006.1 | NP_001159478.1 | 6363263 | c.1039G>T | p.V347L | |
7428413 | c.1071G>A | p.P357P | |||
6654560 | c.919C>T | p.L307F | |||
7308789 | c.906-4G>C | p.? | |||
6654558 | c.928C>T | p.R310W | |||
7221653 | c.1002G>T | p.Q334H | |||
6390563 | c.958T>C | p.C320R | |||
6654586 | c.1054G>T | p.D352Y | |||
6364392 | c.925C>A | p.L309I | |||
NM_001166007.1 | NP_001159479.1 | - | - | - | |
NM_004437.3 | NP_004428.1 | - | - | - | |
NM_203342.2 | NP_976217.1 | - | - | - | |
NM_203343.2 | NP_976218.1 | - | - | - | |
XM_005245753.1 | XP_005245810.1 | - | - | - | |
XM_005245757.1 | XP_005245814.1 | - | - | - | |
XM_005245760.1 | XP_005245817.1 | - | - | - | |
XM_005245761.1 | XP_005245818.1 | - | - | - | |
XM_005245763.1 | XP_005245820.1 | - | - | - | |
XM_005245764.1 | XP_005245821.1 | - | - | - | |
XM_005245765.1 | XP_005245822.1 | - | - | - | |
XM_005245768.1 | XP_005245825.1 | - | - | - | |
XM_005245769.1 | XP_005245826.1 | - | - | - | |
XM_005245770.1 | XP_005245827.1 | - | - | - | |
XM_005245772.4 | XP_005245829.1 | - | - | - | |
XM_005245773.4 | XP_005245830.1 | - | - | - | |
XM_005245774.1 | XP_005245831.1 | - | - | - | |
XM_006710434.1 | XP_006710497.1 | - | - | - | |
XM_006710439.1 | XP_006710502.1 | - | - | - | |
XM_011540956.1 | XP_011539258.1 | - | - | - | |
XM_011540957.1 | XP_011539259.1 | - | - | - | |
XM_011540958.1 | XP_011539260.1 | - | - | - | |
XM_011540959.1 | XP_011539261.1 | - | - | - | |
XM_011540960.1 | XP_011539262.1 | - | - | - | |
XM_011540961.1 | XP_011539263.1 | - | - | - | |
XM_011540962.1 | XP_011539264.1 | - | - | - | |
XM_011540963.2 | XP_011539265.1 | - | - | - | |
XM_011540964.1 | XP_011539266.1 | - | - | - | |
XM_011540965.2 | XP_011539267.1 | - | - | - | |
XM_017000581.1 | XP_016856070.1 | - | - | - | |
XM_017000582.1 | XP_016856071.1 | - | - | - | |
XM_017000583.1 | XP_016856072.1 | - | - | - | |
XM_017000584.1 | XP_016856073.1 | - | - | - | |
XM_017000585.1 | XP_016856074.1 | - | - | - | |
XM_017000586.1 | XP_016856075.1 | - | - | - | |
XM_017000587.1 | XP_016856076.1 | - | - | - | |
XM_017000588.1 | XP_016856077.1 | - | - | - | |
XM_017000589.1 | XP_016856078.1 | - | - | - | |
XM_017000590.1 | XP_016856079.1 | - | - | - | |
XM_017000591.1 | XP_016856080.1 | - | - | - | |
XM_017000592.1 | XP_016856081.1 | - | - | - | |
XM_017000593.1 | XP_016856082.1 | - | - | - | |
XM_017000594.1 | XP_016856083.1 | - | - | - | |
XM_017000595.1 | XP_016856084.1 | 1251241 | c.372C>T | p.I124I | |
7428414 | c.444G>A | p.P148P | |||
426042 | c.443C>G | p.P148R | |||
7308790 | c.279-4G>C | p.? | |||
2152926 | c.394G>A | p.D132N | |||
5214503 | c.429delT | p.F144fs*25 | |||
6390564 | c.331T>C | p.C111R | |||
6364393 | c.298C>A | p.L100I | |||
4977761 | c.375G>T | p.Q125H | |||
6363264 | c.412G>T | p.V138L | |||
4681159 | c.292C>T | p.L98F | |||
6654587 | c.427G>T | p.D143Y | |||
5993153 | c.411C>T | p.G137G | |||
908013 | c.400G>T | p.E134* | |||
6654559 | c.301C>T | p.R101W | |||
XM_017000596.1 | XP_016856085.1 | - | - | - | |
XM_017000597.1 | XP_016856086.1 | - | - | - | |
XM_017000598.1 | XP_016856087.1 | - | - | - | |
XM_017000599.1 | XP_016856088.1 | - | - | - | |
XM_017000600.1 | XP_016856089.1 | - | - | - | |
XM_017000601.1 | XP_016856090.1 | - | - | - | |
XM_017000602.1 | XP_016856091.1 | - | - | - | |
XM_017000603.1 | XP_016856092.1 | - | - | - | |
XM_017000604.1 | XP_016856093.1 | - | - | - | |
GenBank | AB209649.1 | BAD92886.1 | - | - | - |
AF156225.1 | AAD42222.1 | - | - | - | |
AK310655.1 | - | - | - | ||
BC039079.1 | AAH39079.1 | - | - | - | |
BC096104.1 | AAH96104.1 | - | - | - | |
BC096105.1 | - | - | - | ||
BC113841.1 | AAI13842.1 | - | - | - | |
BC114450.1 | AAI14451.1 | - | - | - | |
ENST00000349460.0 | - | - | - | ||
J03796.1 | AAA35794.1 | - | - | - | |
M14993.1 | AAA35795.1 | - | - | - | |
M61733.1 | AAA35797.1 | - | - | - |
Literature Links: |
EPB41 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |