Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
DTNA | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001128175.1 | NP_001121647.1 | 5854123 | c.92G>A | p.R31Q |
4153962 | c.104A>T | p.Y35F | |||
3821378 | c.135G>C | p.Q45H | |||
1290822 | c.109A>G | p.T37A | |||
5854130 | c.94C>T | p.L32F | |||
5810136 | c.103T>A | p.Y35N | |||
4071951 | c.144C>T | p.C48C | |||
1589460 | c.91C>T | p.R31* | |||
3821385 | c.138G>T | p.K46N | |||
NM_001198938.1 | NP_001185867.1 | - | - | - | |
NM_001198939.1 | NP_001185868.1 | 5854120 | c.92G>A | p.R31Q | |
4071947 | c.144C>T | p.C48C | |||
3821381 | c.138G>T | p.K46N | |||
5854127 | c.94C>T | p.L32F | |||
4153959 | c.104A>T | p.Y35F | |||
5810133 | c.103T>A | p.Y35N | |||
3821374 | c.135G>C | p.Q45H | |||
NM_001198940.1 | NP_001185869.1 | - | - | - | |
NM_001198941.1 | NP_001185870.1 | - | - | - | |
NM_001198945.1 | NP_001185874.1 | - | - | - | |
NM_001390.4 | NP_001381.2 | 1290820 | c.109A>G | p.T37A | |
5810132 | c.103T>A | p.Y35N | |||
4071946 | c.144C>T | p.C48C | |||
3821380 | c.138G>T | p.K46N | |||
5854126 | c.94C>T | p.L32F | |||
4153958 | c.104A>T | p.Y35F | |||
988035 | c.91C>T | p.R31* | |||
3821373 | c.135G>C | p.Q45H | |||
5854119 | c.92G>A | p.R31Q | |||
NM_001391.5 | NP_001382.2 | - | - | - | |
NM_001392.4 | NP_001383.2 | 4071949 | c.144C>T | p.C48C | |
5810135 | c.103T>A | p.Y35N | |||
5854122 | c.92G>A | p.R31Q | |||
3821376 | c.135G>C | p.Q45H | |||
1290821 | c.109A>G | p.T37A | |||
1589459 | c.91C>T | p.R31* | |||
4153961 | c.104A>T | p.Y35F | |||
5854129 | c.94C>T | p.L32F | |||
3821383 | c.138G>T | p.K46N | |||
NM_032975.3 | NP_116757.2 | - | - | - | |
NM_032978.6 | NP_116760.2 | - | - | - | |
NM_032979.4 | NP_116761.2 | - | - | - | |
XM_005258221.4 | XP_005258278.1 | - | - | - | |
XM_011525853.2 | XP_011524155.1 | - | - | - | |
XM_017025575.1 | XP_016881064.1 | - | - | - | |
XM_017025576.1 | XP_016881065.1 | - | - | - | |
XM_017025577.1 | XP_016881066.1 | - | - | - | |
XM_017025578.1 | XP_016881067.1 | - | - | - | |
XM_017025579.1 | XP_016881068.1 | - | - | - | |
XM_017025580.1 | XP_016881069.1 | - | - | - | |
XM_017025581.1 | XP_016881070.1 | - | - | - | |
XM_017025582.1 | XP_016881071.1 | - | - | - | |
XM_017025583.1 | XP_016881072.1 | - | - | - | |
XM_017025584.1 | XP_016881073.1 | - | - | - | |
XM_017025585.1 | XP_016881074.1 | - | - | - | |
XM_017025586.1 | XP_016881075.1 | - | - | - | |
XM_017025587.1 | XP_016881076.1 | - | - | - | |
XM_017025588.1 | XP_016881077.1 | - | - | - | |
XM_017025589.1 | XP_016881078.1 | - | - | - | |
XM_017025590.1 | XP_016881079.1 | - | - | - | |
XM_017025591.1 | XP_016881080.1 | - | - | - | |
XM_017025592.1 | XP_016881081.1 | - | - | - | |
XM_017025593.1 | XP_016881082.1 | - | - | - | |
XM_017025594.1 | XP_016881083.1 | - | - | - | |
XM_017025595.1 | XP_016881084.1 | - | - | - | |
XM_017025596.1 | XP_016881085.1 | - | - | - | |
XM_017025597.1 | XP_016881086.1 | - | - | - | |
XM_017025598.1 | XP_016881087.1 | - | - | - | |
XM_017025600.1 | XP_016881089.1 | - | - | - | |
XM_017025601.1 | XP_016881090.1 | - | - | - | |
XM_017025602.1 | XP_016881091.1 | - | - | - | |
GenBank | AB209102.1 | BAD92339.1 | - | - | - |
AJ009668.1 | CAA08769.1 | - | - | - | |
AK291156.1 | - | - | - | ||
AK294746.1 | - | - | - | ||
AK308410.1 | - | - | - | ||
AL833285.1 | - | - | - | ||
BC005300.1 | AAH05300.1 | - | - | - | |
BT006937.1 | AAP35583.1 | - | - | - | |
DA073612.1 | - | - | - | ||
DA197970.1 | - | - | - | ||
DA398530.1 | - | - | - | ||
U26742.1 | AAC50424.1 | - | - | - | |
U26744.1 | AAC50426.1 | - | - | - | |
U46744.1 | AAC50429.1 | - | - | - | |
U46745.1 | AAC50430.1 | - | - | - |
Literature Links: |
DTNA PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |