Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CFLAR | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001127183.2 | NP_001120655.1 | - | - | - |
NM_001127184.2 | NP_001120656.1 | 1306293 | c.409G>A | p.E137K | |
3576253 | c.501A>T | p.K167N | |||
6543801 | c.492G>T | p.L164L | |||
3576250 | c.442C>T | p.Q148* | |||
6987865 | c.507G>T | p.Q169H | |||
6849220 | c.416A>G | p.E139G | |||
6627569 | c.510G>A | p.K170K | |||
NM_001202516.1 | NP_001189445.1 | - | - | - | |
NM_001202517.1 | NP_001189446.1 | - | - | - | |
NM_001202518.1 | NP_001189447.1 | - | - | - | |
NM_001202519.1 | NP_001189448.1 | - | - | - | |
NM_001308042.1 | NP_001294971.1 | - | - | - | |
NM_001308043.1 | NP_001294972.1 | - | - | - | |
NM_003879.5 | NP_003870.4 | 6987864 | c.507G>T | p.Q169H | |
6849219 | c.416A>G | p.E139G | |||
3576249 | c.442C>T | p.Q148* | |||
1306292 | c.409G>A | p.E137K | |||
6543800 | c.492G>T | p.L164L | |||
3576252 | c.501A>T | p.K167N | |||
6627568 | c.510G>A | p.K170K | |||
XM_017005189.1 | XP_016860678.1 | - | - | - | |
XM_017005190.1 | XP_016860679.1 | - | - | - | |
XM_017005191.1 | XP_016860680.1 | - | - | - | |
XM_017005192.1 | XP_016860681.1 | - | - | - | |
XM_017005193.1 | XP_016860682.1 | - | - | - | |
XM_017005194.1 | XP_016860683.1 | - | - | - | |
XM_017005195.1 | XP_016860684.1 | - | - | - | |
XM_017005196.1 | XP_016860685.1 | - | - | - | |
XM_017005197.1 | XP_016860686.1 | - | - | - | |
XM_017005198.1 | XP_016860687.1 | - | - | - | |
GenBank | AB209600.1 | BAD92837.1 | - | - | - |
AF005774.1 | AAC15825.1 | - | - | - | |
AF005775.1 | AAC15826.1 | - | - | - | |
AF009616.1 | AAB70909.1 | - | - | - | |
AF009617.1 | AAB70910.1 | - | - | - | |
AF009618.1 | AAB70911.1 | - | - | - | |
AF009619.1 | AAB70912.1 | - | - | - | |
AF010127.1 | AAB64110.1 | - | - | - | |
AF015450.1 | AAC16439.1 | - | - | - | |
AF015451.1 | AAC16440.1 | - | - | - | |
AF015452.1 | AAC16441.1 | - | - | - | |
AF041458.1 | AAB99790.1 | - | - | - | |
AF041460.1 | AAB99792.1 | - | - | - | |
AF041461.1 | AAB99793.1 | - | - | - | |
AF041462.1 | AAB99794.1 | - | - | - | |
AK289913.1 | - | - | - | ||
AK296036.1 | - | - | - | ||
AK297387.1 | - | - | - | ||
AK304585.1 | - | - | - | ||
AK308767.1 | - | - | - | ||
AK315208.1 | - | - | - | ||
AK315924.1 | - | - | - | ||
BC001602.1 | AAH01602.1 | - | - | - | |
BT006751.1 | AAP35397.1 | - | - | - | |
CD699251.1 | - | - | - | ||
ENST00000309955.0 | - | - | - | ||
U85059.1 | AAB82648.1 | - | - | - | |
U97074.1 | AAC51622.1 | - | - | - | |
U97075.1 | AAC51623.1 | - | - | - | |
Y14039.1 | CAA74366.1 | - | - | - | |
Y14040.1 | CAA74367.1 | - | - | - |
Literature Links: |
CFLAR PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |