Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACNA1G | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001256324.1 | NP_001243253.1 | - | - | - |
NM_001256325.1 | NP_001243254.1 | - | - | - | |
NM_001256326.1 | NP_001243255.1 | - | - | - | |
NM_001256327.1 | NP_001243256.1 | - | - | - | |
NM_001256328.1 | NP_001243257.1 | - | - | - | |
NM_001256329.1 | NP_001243258.1 | - | - | - | |
NM_001256330.1 | NP_001243259.1 | - | - | - | |
NM_001256331.1 | NP_001243260.1 | - | - | - | |
NM_001256332.1 | NP_001243261.1 | - | - | - | |
NM_001256333.1 | NP_001243262.1 | - | - | - | |
NM_001256334.1 | NP_001243263.1 | - | - | - | |
NM_001256359.1 | NP_001243288.1 | - | - | - | |
NM_001256360.1 | NP_001243289.1 | - | - | - | |
NM_001256361.1 | NP_001243290.1 | - | - | - | |
NM_018896.4 | NP_061496.2 | 6532498 | c.1303G>C | p.E435Q | |
473015 | c.1185C>T | p.A395A | |||
1384290 | c.1244G>A | p.R415Q | |||
3356937 | c.1333C>T | p.R445C | |||
1630273 | c.1330C>G | p.L444V | |||
4831305 | c.1311C>G | p.L437L | |||
3192180 | c.1335T>C | p.R445R | |||
5386703 | c.1148C>T | p.S383F | |||
3742314 | c.1189C>G | p.Q397E | |||
1384286 | c.1188G>A | p.T396T | |||
5386711 | c.1222C>T | p.L408L | |||
1630269 | c.1288G>A | p.G430S | |||
3691685 | c.1209G>A | p.Q403Q | |||
3192172 | c.1238G>A | p.R413H | |||
3519208 | c.1173G>A | p.L391L | |||
3889921 | c.1155C>T | p.F385F | |||
5386707 | c.1212G>A | p.R404R | |||
1199305 | c.1156A>G | p.M386V | |||
4067685 | c.1296C>A | p.C432* | |||
981141 | c.1319T>C | p.L440P | |||
NM_198376.2 | NP_938190.1 | - | - | - | |
NM_198377.2 | NP_938191.2 | - | - | - | |
NM_198378.2 | NP_938192.1 | - | - | - | |
NM_198379.2 | NP_938193.1 | - | - | - | |
NM_198380.2 | NP_938194.1 | - | - | - | |
NM_198382.2 | NP_938196.1 | - | - | - | |
NM_198383.2 | NP_938197.1 | - | - | - | |
NM_198384.2 | NP_938198.1 | - | - | - | |
NM_198385.2 | NP_938199.1 | - | - | - | |
NM_198386.2 | NP_938200.1 | - | - | - | |
NM_198387.2 | NP_938201.1 | - | - | - | |
NM_198388.2 | NP_938202.1 | - | - | - | |
NM_198396.2 | NP_938406.1 | 3742312 | c.1189C>G | p.Q397E | |
5386705 | c.1212G>A | p.R404R | |||
4067683 | c.1296C>A | p.C432* | |||
3691683 | c.1209G>A | p.Q403Q | |||
1384284 | c.1188G>A | p.T396T | |||
1199303 | c.1156A>G | p.M386V | |||
1630267 | c.1288G>A | p.G430S | |||
3192170 | c.1238G>A | p.R413H | |||
3519206 | c.1173G>A | p.L391L | |||
6532496 | c.1303G>C | p.E435Q | |||
5386701 | c.1148C>T | p.S383F | |||
5386709 | c.1222C>T | p.L408L | |||
981139 | c.1319T>C | p.L440P | |||
473013 | c.1185C>T | p.A395A | |||
1384288 | c.1244G>A | p.R415Q | |||
3356935 | c.1333C>T | p.R445C | |||
4831303 | c.1311C>G | p.L437L | |||
1630271 | c.1330C>G | p.L444V | |||
3192178 | c.1335T>C | p.R445R | |||
3889919 | c.1155C>T | p.F385F | |||
NR_046054.1 | - | - | - | ||
NR_046055.1 | - | - | - | ||
NR_046056.1 | - | - | - | ||
NR_046057.1 | - | - | - | ||
NR_046058.1 | - | - | - | ||
XM_006722160.3 | XP_006722223.1 | - | - | - | |
XM_006722161.3 | XP_006722224.1 | - | - | - | |
GenBank | AB012043.1 | BAA36409.1 | - | - | - |
AF124351.1 | AAD34352.1 | - | - | - | |
AF126965.1 | AAD29400.1 | - | - | - | |
AF126966.1 | AAD29401.1 | - | - | - | |
AF134985.1 | AAF19346.1 | - | - | - | |
AF134986.1 | AAF19347.1 | - | - | - | |
AF190860.1 | AAF35287.1 | - | - | - | |
AF227744.1 | AAF37689.1 | - | - | - | |
AF227745.1 | AAF37690.1 | - | - | - | |
AF227746.1 | AAF37691.1 | - | - | - | |
AF227747.1 | AAF37692.1 | - | - | - | |
AF227748.1 | AAF37693.1 | - | - | - | |
AF227749.1 | AAF37694.1 | - | - | - | |
AF227750.1 | AAF37695.1 | - | - | - | |
AF227751.1 | AAF37696.1 | - | - | - | |
BC110995.1 | AAI10996.1 | - | - | - | |
DQ494449.1 | - | - | - | ||
DQ494450.1 | - | - | - | ||
DQ494451.1 | - | - | - | ||
DQ494452.1 | - | - | - | ||
DQ494453.1 | - | - | - | ||
DQ494454.1 | - | - | - | ||
DQ494455.1 | - | - | - | ||
DQ494456.1 | - | - | - | ||
DQ494457.1 | - | - | - | ||
DQ494458.1 | - | - | - | ||
DQ494459.1 | - | - | - | ||
DQ494460.1 | - | - | - | ||
DQ494461.1 | - | - | - | ||
DQ494462.1 | - | - | - | ||
DQ494463.1 | - | - | - | ||
DQ494464.1 | - | - | - | ||
DQ494465.1 | - | - | - | ||
DQ494466.1 | - | - | - | ||
DQ494467.1 | - | - | - | ||
DQ494468.1 | - | - | - | ||
DQ494469.1 | - | - | - | ||
DQ494470.1 | - | - | - | ||
DQ494471.1 | - | - | - | ||
DQ494472.1 | - | - | - | ||
DQ494473.1 | - | - | - | ||
DQ494474.1 | - | - | - | ||
DQ494475.1 | - | - | - | ||
DQ494476.1 | - | - | - | ||
DQ494477.1 | - | - | - | ||
DQ494478.1 | - | - | - | ||
DQ494479.1 | - | - | - | ||
KJ534993.1 | - | - | - | ||
KJ535064.1 | - | - | - |
Literature Links: |
CACNA1G PubMed Links |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
Set Membership: |
Between 200-400 bp |