Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ESRRG | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001134285.2 | NP_001127757.1 | - | - | - |
NM_001243505.1 | NP_001230434.1 | - | - | - | |
NM_001243506.1 | NP_001230435.1 | - | - | - | |
NM_001243507.1 | NP_001230436.1 | - | - | - | |
NM_001243509.1 | NP_001230438.1 | - | - | - | |
NM_001243510.2 | NP_001230439.1 | - | - | - | |
NM_001243511.2 | NP_001230440.1 | - | - | - | |
NM_001243512.1 | NP_001230441.1 | - | - | - | |
NM_001243513.1 | NP_001230442.1 | - | - | - | |
NM_001243514.1 | NP_001230443.1 | - | - | - | |
NM_001243515.1 | NP_001230444.1 | 7213519 | c.1230G>C | p.Q410H | |
6231500 | c.1173C>A | p.H391Q | |||
4187361 | c.1269A>C | p.K423N | |||
5590831 | c.1312T>C | p.L438L | |||
3483702 | c.1186C>T | p.R396C | |||
3483699 | c.1221C>T | p.L407L | |||
4864152 | c.1209G>T | p.M403I | |||
4028251 | c.1187G>A | p.R396H | |||
7246344 | c.1259A>T | p.Y420F | |||
5716380 | c.1182C>T | p.D394D | |||
3864512 | c.1177G>A | p.E393K | |||
3803906 | c.1206G>A | p.L402L | |||
7213466 | c.1273G>A | p.E425K | |||
6994637 | c.1290G>T | p.M430I | |||
4784964 | c.1322A>C | p.K441T | |||
4872313 | c.1280A>G | p.K427R | |||
3483701 | c.1189C>T | p.R397* | |||
6656510 | c.1296A>C | p.K432N | |||
3483697 | c.1296A>G | p.K432K | |||
6656506 | c.1300T>G | p.F434V | |||
4028249 | c.1322A>G | p.K441R | |||
4681772 | c.1246G>A | p.V416M | |||
7287158 | c.1308A>G | p.E436E | |||
4612866 | c.1304delT | p.L435fs*>8 | |||
5776637 | c.1268A>C | p.K423T | |||
7246946 | c.1238C>A | p.T413N | |||
NM_001243518.1 | NP_001230447.1 | - | - | - | |
NM_001243519.1 | NP_001230448.1 | - | - | - | |
NM_001438.3 | NP_001429.2 | 374752 | c.1371G>T | p.K457N | |
7246343 | c.1307A>T | p.Y436F | |||
361445 | c.1238G>C | p.R413P | |||
7213465 | c.1321G>A | p.E441K | |||
6994636 | c.1338G>T | p.M446I | |||
5716379 | c.1230C>T | p.D410D | |||
3483700 | c.1237C>T | p.R413* | |||
903964 | c.1257G>T | p.M419I | |||
4028248 | c.1370A>G | p.K457R | |||
3483696 | c.1344A>G | p.K448K | |||
1958519 | c.1234C>T | p.R412C | |||
6231499 | c.1221C>A | p.H407Q | |||
3864511 | c.1225G>A | p.E409K | |||
381773 | c.1371G>C | p.K457N | |||
5590830 | c.1360T>C | p.L454L | |||
334251 | c.1231C>A | p.P411T | |||
3803905 | c.1254G>A | p.L418L | |||
7246945 | c.1286C>A | p.T429N | |||
4187360 | c.1317A>C | p.K439N | |||
6656509 | c.1344A>C | p.K448N | |||
7213518 | c.1278G>C | p.Q426H | |||
1338888 | c.1370A>C | p.K457T | |||
6656505 | c.1348T>G | p.F450V | |||
5776636 | c.1316A>C | p.K439T | |||
4028250 | c.1235G>A | p.R412H | |||
903963 | c.1328A>G | p.K443R | |||
3483698 | c.1269C>T | p.L423L | |||
386226 | c.1301A>T | p.H434L | |||
4681771 | c.1294G>A | p.V432M | |||
7287157 | c.1356A>G | p.E452E | |||
4612865 | c.1352delT | p.L451fs*>8 | |||
NM_206594.2 | NP_996317.1 | - | - | - | |
NM_206595.2 | NP_996318.1 | - | - | - | |
XM_011509265.2 | XP_011507567.1 | - | - | - | |
XM_011509266.2 | XP_011507568.1 | - | - | - | |
XM_011509267.1 | XP_011507569.1 | - | - | - | |
XM_011509268.2 | XP_011507570.1 | - | - | - | |
XM_011509269.2 | XP_011507571.1 | - | - | - | |
XM_011509270.1 | XP_011507572.1 | - | - | - | |
XM_011509271.2 | XP_011507573.1 | - | - | - | |
XM_011509274.1 | XP_011507576.1 | - | - | - | |
XM_011509275.1 | XP_011507577.1 | - | - | - | |
XM_011509276.1 | XP_011507578.1 | - | - | - | |
XM_011509277.1 | XP_011507579.1 | - | - | - | |
XM_011509278.1 | XP_011507580.1 | - | - | - | |
XM_011509279.1 | XP_011507581.1 | - | - | - | |
XM_011509280.2 | XP_011507582.1 | - | - | - | |
XM_017000621.1 | XP_016856110.1 | - | - | - | |
XM_017000622.1 | XP_016856111.1 | - | - | - | |
XM_017000623.1 | XP_016856112.1 | - | - | - | |
XM_017000624.1 | XP_016856113.1 | - | - | - | |
XM_017000625.1 | XP_016856114.1 | - | - | - | |
XM_017000626.1 | XP_016856115.1 | - | - | - | |
XM_017000627.1 | XP_016856116.1 | - | - | - | |
XM_017000628.1 | XP_016856117.1 | - | - | - | |
XM_017000629.1 | XP_016856118.1 | - | - | - | |
XM_017000630.1 | XP_016856119.1 | - | - | - | |
XM_017000631.1 | XP_016856120.1 | - | - | - | |
XM_017000632.1 | XP_016856121.1 | - | - | - | |
XM_017000633.1 | XP_016856122.1 | - | - | - | |
XM_017000634.1 | XP_016856123.1 | - | - | - | |
XM_017000635.1 | XP_016856124.1 | - | - | - | |
XM_017000636.1 | XP_016856125.1 | - | - | - | |
XM_017000637.1 | XP_016856126.1 | - | - | - | |
XM_017000638.1 | XP_016856127.1 | - | - | - | |
XM_017000639.1 | XP_016856128.1 | - | - | - | |
XM_017000640.1 | XP_016856129.1 | - | - | - | |
XM_017000641.1 | XP_016856130.1 | - | - | - | |
XM_017000642.1 | XP_016856131.1 | - | - | - | |
XM_017000643.1 | XP_016856132.1 | - | - | - | |
XM_017000644.1 | XP_016856133.1 | - | - | - | |
XM_017000645.1 | XP_016856134.1 | - | - | - | |
XM_017000646.1 | XP_016856135.1 | - | - | - | |
XM_017000647.1 | XP_016856136.1 | - | - | - | |
XM_017000648.1 | XP_016856137.1 | - | - | - | |
XM_017000649.1 | XP_016856138.1 | - | - | - | |
GenBank | AB020639.1 | - | - | - | |
AB307715.1 | - | - | - | ||
AB362218.1 | - | - | - | ||
AF058291.1 | AAC39899.1 | - | - | - | |
AF094518.1 | AAC99410.1 | - | - | - | |
AK131193.1 | - | - | - | ||
AK290945.1 | - | - | - | ||
AK291028.1 | - | - | - | ||
AK291647.1 | - | - | - | ||
AK298851.1 | - | - | - | ||
AY388456.1 | AAQ93376.1 | - | - | - | |
AY388457.1 | AAQ93377.1 | - | - | - | |
AY388458.1 | AAQ93378.1 | - | - | - | |
AY388459.1 | AAQ93379.1 | - | - | - | |
AY388460.1 | AAQ93380.1 | - | - | - | |
AY388461.1 | AAQ93381.1 | - | - | - | |
BC064700.1 | AAH64700.1 | - | - | - | |
BX486063.1 | - | - | - | ||
BX647076.1 | - | - | - | ||
CR749497.1 | CAH18320.1 | - | - | - | |
ENST00000408911.0 | - | - | - | ||
HQ692853.1 | - | - | - | ||
HQ692854.1 | - | - | - |
Literature Links: |
ESRRG PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |