Search
The Arcturus® Paradise® PLUS FFPE RNA Isolation Kit enables unprecedented gene expression analysis of millions of previously inaccessible samples by unlocking RNA from formalin-fixed, paraffin-embedded (FFPE) tissues. Amplify as little as 5 ng of the isolated fixed RNA using reagents and methods that have been specially optimized to overcome the challenges of cross-linked FFPE templates (Figure 1).
![]() | Figure 1. Samples processed using the Paradise® PLUS Reagent System. Starting with 5,000–10,000 laser capture microdissected cells produces amplified RNA sufficient for duplicate microarray hybridizations. |
Not all FFPE samples contain high-quality RNA. Identify the best samples for your studies using a simple real-time PCR research technique, and then proceed with confidence using these prequalified samples (Figure 2).
| Figure 2. Process for Paradise® PLUS isolation provides a simple path to qPCR success. |
The Paradise® PLUS Reagent Systems is validated as part of the Arcturus® System for Microgenomics®, an integrated solution for utilizing small quantities of RNA for gene expression analysis, and features Arcturus Laser Capture Microdissection (LCM) instruments to prepare pure cell populations.
For Research Use Only. Not for use in diagnostic procedures.