Search
We are dedicated to your success. Get back on track. View our expert recommendations for commonly encountered problem scenarios.
View the relevant questions below:
There are two things that affect a DNA band’s migration as it travels through the gel matrix.
Many whitening agents used in clothing, as well as some fungi and bacteria, fluoresce at the same wavelengths as SYBR® Safe DNA gel stain. These contaminants within or on the surface of the gel may produce this speckling.
This can happen if the marker was heated. Please ensure that the ladders are not heated before use.
Here are a few reasons why you might see smearing of the bands:
There are a few possible reasons for this:
For Research Use Only. Not for use in diagnostic procedures.