Fast, trusted COVID-19 test results from raw saliva ideal for widespread, high frequency testing
Features of the TaqPath COVID-19 Fast PCR Combo Kit 2.0:
- Fast, direct-to-PCR workflow from raw saliva—increased speed from raw saliva samples to results, enabling widespread, high-frequency testing
- Simple and affordable—go from raw saliva collection direct to PCR (no RNA extraction), helping save supplies, reagents, and personnel costs
- Gold standard technology—a sensitive, PCR-based test offering a limit of detection of 750–1,000 GCE/mL from raw saliva
- Innovative, multi-target design—multiple targets on orf1a, orf1b, and N genes compensate for emerging SARS-CoV-2 mutations, helping provide increased confidence in results
Fast, direct-to-PCR workflow
The TaqPath COVID-19 Fast PCR Combo Kit 2.0 workflow does not require RNA extraction and provides results in ~2 hours.
Receive samples | 
Prepare samples | 
Bypass extraction | 
Prepare plate and run PCR | 
Analyze and report |
Raw saliva (no preservatives) |
Lysis solution
| - TaqPath COVID-19 Fast PCR Combo Kit 2.0
- Real-time PCR instruments:
- QuantStudio 5 System (96-well, 0.2 mL)
- QuantStudio 7 System (384-well)
| | Applied Biosystems Pathogen Interpretive Software (v1.1.0) |
Advantages of saliva collection [1]- Recent studies have shown no statistically significant difference in sensitivity between nasopharyngeal swab (NPS) and saliva collection for SARS-CoV-2 detection
- Saliva is easily self-collected, reducing both the exposure to health care providers and the need for personal protective equipment (PPE)
- When including the cost of swabs, transport media, containers, PPE, and labor costs, saliva collection can save significant amounts of money compared to using NPS for SARS-CoV-2 detection
Find your IFU
Find the approved Instructions for Use (IFU) for your country. (Note: Product availability varies by country.)
Find out more
| Product | Component | Qty | Cat. No. |
| TaqPath COVID-19 Fast PCR Combo Kit 2.0 | TaqPath COVID-19 Fast PCR Assay 2.0 (1 tube) TaqPath COVID-19 Plus Control (10 tubes) TaqPath Control Dilution Buffer (1 box with 10 tubes) SalivaReady Solution (1 bottle with 20 mL) TaqPath 1-Step Multiplex Master Mix, No ROX (1 bottle with 10 mL) (required, but not included in kit) | 1,000 rxn | A51606 |
Simple analysis and reporting
Applied Biosystems Pathogen Interpretive Software (v1.1.0) is part of the required workflow and must be downloaded prior to instrumentation use.
Pathogen Interpretive Software helps your lab decrease analysis and interpretation time and reduce risk of user interpretation error. You may download the software upon kit purchase. Labs following the authorized workflow are required to complete an eLearning course and pass an exam to download.
Follow the instructions below to obtain the software from your local support team:
- Go to thermofisher.com/contactus
- Select “Technical Support” as category
- Enter product name or SKU and select your country
- Call the local number displayed on the screen
Reference
1. Bastos, M. et al. (2021) The Sensitivity and Costs of Testing for SARS-CoV-2 Infection With Saliva Versus Nasopharyngeal Swabs. Annals of Internal Medicine. 174(4):501-510.
For In Vitro Diagnostic Use. For Emergency Use Authorization (EUA) Only | Rx Only
© 2021 Thermo Fisher Scientific, Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified